Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01146147_m1
          See other NDE1 GE Assays ›
          Gene Symbol:
          Nde1
          Gene Name:
          nuclear distribution gene E homolog 1 (A nidulans)
          Gene Aliases:
          2810027M15Rik, AU042936, AW822251, Nude, mNudE
          Chromosome Location:
          Chr.16: 14163275 - 14192928 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01146147_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nde1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001285504.1 NP_001272433.1 7 - 8 933 63
          NM_023317.2 NP_075806.2 8 - 9 1082 63
          GenBank mRNA AF290473.1 - 6 - 7 816 63
          AF322073.1 - 7 - 8 989 63
          AK012830.1 - 8 - 9 1012 63
          AK012861.1 - 8 - 9 1030 63
          AK037289.1 - 8 - 9 1025 63
          AK166858.1 - 7 - 8 912 63
          AK169743.1 - 8 - 9 1012 63
          BC023267.1 - 8 - 9 1061 3672565 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000176.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nde1

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          mitotic cell cycle
          neuron migration
          microtubule nucleation
          cell cycle
          chromosome segregation
          mitotic centrosome separation
          multicellular organism development
          nervous system development
          neuroblast proliferation
          cerebral cortex development
          cell differentiation
          forebrain development
          microtubule organizing center organization
          vesicle transport along microtubule
          centrosome duplication
          cell division
          establishment of chromosome localization
          centrosome localization
          protein binding
          microtubule binding
          protein domain specific binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600001272.mm.1 TC1600000176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001272.mm.1 NM_001285503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001272.mm.1 NM_001285504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001272.mm.1 NM_023317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001272.mm.1 NM_023317.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 TC1600000176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 NM_001114085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 NM_001114085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 NM_001285503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 NM_001285504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 NM_023317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001273.mm.1 NM_023317.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 TC1600000176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 NM_001114085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 NM_001114085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 NM_001285503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 NM_001285504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 NM_023317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001275.mm.1 NM_023317.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0