Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01137867_m1
          See other CTBP2 GE Assays ›
          Gene Symbol:
          Ctbp2
          Gene Name:
          C-terminal binding protein 2
          Gene Aliases:
          AA407280, D7Ertd45e, Gtrgeo6, Ribeye
          Chromosome Location:
          Chr.7: 132986417 - 133123877 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01137867_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009980.4 NP_034110.1 1 - 2 340 66
          GenBank mRNA AB033123.1 - 1 - 2 205 66
          AK146713.1 - 1 - 2 225 66
          AK147871.1 - 1 - 2 223 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004667.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB033123.1, AK146713.1, AK147871.1, NM_009980.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctbp2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          metabolic process
          cell differentiation
          positive regulation of chromatin binding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          positive regulation of retinoic acid receptor signaling pathway
          white fat cell differentiation
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          retinoic acid receptor binding
          NAD binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700036902.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036902.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036902.mm.1 NM_009980.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036899.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036899.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036899.mm.1 NM_009980.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036901.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036901.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036901.mm.1 NM_009980.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036907.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036907.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036907.mm.1 NM_009980.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036886.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036886.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036886.mm.1 NM_009980.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036893.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036893.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036893.mm.1 NM_009980.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036908.mm.1 TC0700004667.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036908.mm.1 NM_009980.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036908.mm.1 NM_009980.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0