Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081678.1 | NP_001075147.1 | 4 - 5 | 478 | 74 | |
GenBank mRNA | AK036691.1 | - | 5 - 6 | 744 | 74 | |
BC066073.1 | - | 4 - 5 | 890 | 74 | ||
BC119236.1 | - | 4 - 5 | 478 | 8733985 | 74 | |
BC119262.1 | - | 4 - 5 | 478 | 8734011 | 74 | |
BC144761.1 | - | 4 - 5 | 478 | 9053309 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |