Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005520.2 | NP_001005520.2 | 1 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |