Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838433_g1
          See other STK-PS2 GE Assays ›
          Gene Symbol:
          Stk-ps2
          Gene Name:
          serine/threonine kinase 2
          Gene Aliases:
          4930509O22, EG212225, Gm4776
          Chromosome Location:
          Chr.1: 46020415 - 46030437 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838433_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stk-ps2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037691.1 2 - 3 1758 143
          GenBank mRNA AK029722.1 - 3 - 4 2600 143
          BC145242.1 - 2 - 3 1758 9053794 143
          BC150733.1 - 2 - 3 1758 9087644 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002287.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 BC150733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 BC145241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 BC145242.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 NR_037690.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 NR_037691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 BC150733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 BC145241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 BC145242.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 NR_037690.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498274 NR_037691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 BC150733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 BC145241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 BC145242.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 NR_037690.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 NR_037691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 BC150733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 BC145241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 BC145242.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 NR_037690.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498274 NR_037691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 BC150733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 BC145241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 BC145242.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 NR_037690.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 NR_037691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 BC150733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 BC145241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 BC145242.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 NR_037690.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498274 NR_037691.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00518655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00002771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20199995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00518668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00835959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00970686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00760313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00518686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00970684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00002776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00518667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00835954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20199997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20199994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20200015 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100018454.mm.1 TC0100002287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018454.mm.1 NR_037690.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018454.mm.1 NR_037690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018454.mm.1 NR_037691.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018454.mm.1 NR_037691.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018451.mm.1 TC0100002287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018451.mm.1 NR_037690.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018451.mm.1 NR_037690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018451.mm.1 NR_037691.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018451.mm.1 NR_037691.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018452.mm.1 TC0100002287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018452.mm.1 NR_037691.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018452.mm.1 NR_037691.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 818160 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2628318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2628318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2628318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0