Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835291_m1
          See other ZBTB8OS GE Assays ›
          Gene Symbol:
          Zbtb8os
          Gene Name:
          zinc finger and BTB domain containing 8 opposite strand
          Gene Aliases:
          2010001H09Rik, 2310028N13Rik, AI851226, Arch, Archm
          Chromosome Location:
          Chr.4: 129335560 - 129347083 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835291_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb8os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025970.3 NP_080246.1 2 - 3 139 127
          GenBank mRNA AK007377.1 - 2 - 3 129 127
          AK008007.1 - 2 - 3 140 127
          AK132279.1 - 150 127
          AY071852.1 - 2 - 3 128 127
          BC026442.1 - 134 127
          BC037657.1 - 109 127
          BC046825.1 - 146 127
          BC094629.1 - 2 - 3 170 3376726 127
          BC125606.1 - 2 - 3 140 40130121 127
          BC125608.1 - 2 - 3 140 40130123 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001519.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb8os

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA splicing, via endonucleolytic cleavage and ligation
          tRNA processing
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137152 BC046825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137152 BC094629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137152 BC026442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137152 BC037657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137152 NM_025970.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032238 BC046825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032238 BC094629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032238 BC125606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032238 BC125608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032238 BC026442.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41922 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102882 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163676 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032238 BC046825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032238 BC094629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032238 BC125606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032238 BC125608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032238 BC026442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163344 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137152 BC046825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137152 BC094629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137152 BC026442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137152 BC037657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137152 NM_025970.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147576 BC046825.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147576 BC094629.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147576 BC125606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147576 BC125608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147576 BC026442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176625 BC046825.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176625 BC026442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176625 BC037657.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176625 AK132279.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296230 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163676 - -
          Whole Mouse Genome G4122A A_52_P147576 BC046825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147576 BC094629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147576 BC125606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147576 BC125608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147576 BC026442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176625 BC046825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176625 BC026442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176625 BC037657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176625 AK132279.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137152 BC046825.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137152 BC094629.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137152 BC026442.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137152 BC037657.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137152 NM_025970.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126424 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102882 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163676 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03608557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03344109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03647462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03905877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03463294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03647461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10508445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508448 - -
          Affymetrix HT MG-430 PM Array Plate 1426267_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452006_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1426266_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10508444 - -
          Mouse Genome 430 2.0 Array 1426266_PM_s_at - -
          Mouse Genome 430 2.0 Array 1426267_PM_at - -
          Mouse Genome 430 2.0 Array 1426267_at - -
          Mouse Genome 430 2.0 Array 1452006_PM_x_at - -
          Mouse Genome 430 2.0 Array 1452006_x_at - -
          Mouse Genome 430 2.0 Array 1426266_s_at - -
          Affymetrix Mouse Clariom S Array PSR0400011721.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011721.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011721.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011723.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011723.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011723.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011726.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011726.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011726.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011725.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011725.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011725.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011731.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011720.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011720.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011720.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011729.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011729.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011729.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011722.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011722.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011722.mm.1 NM_025970.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011719.mm.1 TC0400001519.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011719.mm.1 NM_025970.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011719.mm.1 NM_025970.3 Refseq
          Mouse Expression Set 430 1452006_x_at - -
          Mouse Expression Set 430 1452006_PM_x_at - -
          Mouse Expression Set 430 1426266_s_at - -
          Mouse Expression Set 430 1426267_at - -
          Mouse Expression Set 430 1426266_PM_s_at - -
          Mouse Expression Set 430 1426267_PM_at - -
          Murine Genome U74v2 Set 169720_i_at - -
          Murine Genome U74v2 Set 169720_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1452006_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1426267_PM_at - -
          Mouse Genome 430A 2.0 Array 1426267_at - -
          Mouse Genome 430A 2.0 Array 1426266_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452006_x_at - -
          Mouse Genome 430A 2.0 Array 1426266_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452006_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426266_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452006_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426267_at - -
          Affymetrix HT Mouse Genome 430A Array 1426267_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426266_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 830004 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 104120398 - -
          Illumina mouse-6 v1.0 expression beadchip 5690451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740159 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740159 - -
          mouse-6 v1.1 (Illumina) scl067106.7_0-S - -
          mouse-6 v1.1 (Illumina) scl0067106.1_212-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690451 - -
          Illumina Sentrix mouse-6.v1 bead array 104120398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067106.7_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104120398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740159 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2872419 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0