Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833840_g1
          See other COX5B GE Assays ›
          Gene Symbol:
          Cox5b Gm11273
          Gene Name:
          cytochrome c oxidase subunit Vb
          cytochrome c oxidase, subunit Vb pseudogene
          Gene Aliases:
          -
          OTTMUSG00000000426
          Chromosome Location:
          Chr.1: 36691487 - 36693388 on Build GRCm38
          Chromosome Location:
          Chr.13: 21500935 - 21501452 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00833840_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009942.2 NP_034072.2 3 - 4 373 72
          GenBank mRNA AK008339.1 - 3 - 4 294 72
          AK131673.1 - 3 - 4 294 72
          AK135174.1 - 3 - 4 302 72
          AK215114.1 - 1 - 2 78 72
          BC005564.1 - 313 72
          BC096048.1 - 3 - 4 288 3465479 72
          BC099474.1 - 3 - 4 302 30933585 72
          BU757937.1 - 1 - 2 72
          X53157.1 - 3 - 4 282 72
          Gene Symbol: Gm11273
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC023228.1 - 301 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000253.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox5b
          Click here to find siRNAs for Gene Gm11273

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149813 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149812 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149810 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017804 AK131673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017804 AK135174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017804 NM_009942.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017804 AK215114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017804 BC096048.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176528 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155938 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153540 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133695 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129994 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136816 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131068 BC005564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017804 AK131673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017804 AK135174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017804 NM_009942.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017804 AK215114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017804 BC096048.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141123 X53157.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141123 AK008339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141123 AK131673.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141123 AK135174.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141123 NM_009942.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20149823 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153540 - -
          Whole Mouse Genome G4122A A_51_P141123 X53157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141123 AK008339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141123 AK131673.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141123 AK135174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141123 NM_009942.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121736 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109346 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149813 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149812 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20149810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20149807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00440305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00055947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00676055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00676053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10345505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408046 - -
          Affymetrix HT MG-430 PM Array Plate 1436149_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435613_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1456588_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448514_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454716_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1416902_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10345504 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408044 - -
          Mouse Genome 430 2.0 Array 1416902_PM_a_at - -
          Mouse Genome 430 2.0 Array 1456588_x_at - -
          Mouse Genome 430 2.0 Array 1416902_a_at - -
          Mouse Genome 430 2.0 Array 1436149_PM_at - -
          Mouse Genome 430 2.0 Array 1454716_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448514_at - -
          Mouse Genome 430 2.0 Array 1436149_at - -
          Mouse Genome 430 2.0 Array 1435613_x_at - -
          Mouse Genome 430 2.0 Array 1456588_PM_x_at - -
          Mouse Genome 430 2.0 Array 1435613_PM_x_at - -
          Mouse Genome 430 2.0 Array 1454716_x_at - -
          Mouse Genome 430 2.0 Array 1448514_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12859_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12859_at - -
          Affymetrix Mouse Clariom S Array PSR0100001701.mm.1 TC0100000253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001701.mm.1 NM_009942.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001701.mm.1 NM_009942.2 Refseq
          Mouse Expression Set 430 1448514_PM_at - -
          Mouse Expression Set 430 1435613_x_at - -
          Mouse Expression Set 430 1435613_PM_x_at - -
          Mouse Expression Set 430 1456588_PM_x_at - -
          Mouse Expression Set 430 1448514_at - -
          Mouse Expression Set 430 1416902_PM_a_at - -
          Mouse Expression Set 430 1436149_PM_at - -
          Mouse Expression Set 430 1454716_PM_x_at - -
          Mouse Expression Set 430 1456588_x_at - -
          Mouse Expression Set 430 1416902_a_at - -
          Mouse Expression Set 430 1436149_at - -
          Mouse Expression Set 430 1454716_x_at - -
          Murine Genome U74v2 Set 93119_PM_at - -
          Murine Genome U74v2 Set 93119_at - -
          Mouse Genome 430A 2.0 Array 1448514_PM_at - -
          Mouse Genome 430A 2.0 Array 1456588_x_at - -
          Mouse Genome 430A 2.0 Array 1435613_x_at - -
          Mouse Genome 430A 2.0 Array 1436149_at - -
          Mouse Genome 430A 2.0 Array 1416902_a_at - -
          Mouse Genome 430A 2.0 Array 1454716_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1435613_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1456588_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1416902_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1454716_x_at - -
          Mouse Genome 430A 2.0 Array 1436149_PM_at - -
          Mouse Genome 430A 2.0 Array 1448514_at - -
          Affymetrix HT Mouse Genome 430A Array 1448514_at - -
          Affymetrix HT Mouse Genome 430A Array 1448514_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893620 - -
          ABI Mouse Genome Survey Microarray 898865 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105270739 - -
          MouseWG-6 v2 Expression BeadChip 6550670 - -
          Illumina mouse-6 v1.0 expression beadchip 6550670 - -
          Illumina mouse-6 v1.0 expression beadchip 102370563 - -
          Illumina mouse-6 v1.0 expression beadchip 105270739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2721648 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721648 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227230 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772319 - -
          mouse-6 v1.1 (Illumina) scl012859.1_1-S - -
          mouse-6 v1.1 (Illumina) scl0012859.2_116-S - -
          mouse-6 v1.1 (Illumina) scl47494.1.1_88-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550670 - -
          Illumina Sentrix mouse-6.v1 bead array 102370563 - -
          Illumina Sentrix mouse-6.v1 bead array 105270739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0012859.2_116-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105270739 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550670 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102370563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241962 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241962 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772319 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227230 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0012859.2_116-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0