Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00809552_s1
          See other LCN2 GE Assays ›
          Gene Symbol:
          Lcn2
          Gene Name:
          lipocalin 2
          Gene Aliases:
          24p3, AW212229, Sip24
          Chromosome Location:
          Chr.2: 32384637 - 32387739 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00809552_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008491.1 NP_032517.1 6 792 96
          GenBank mRNA AK149774.1 - 6 824 96
          BC020275.1 - 6 792 96
          X14607.1 - 6 792 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003303.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn2

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          transport
          ion transport
          apoptotic process
          response to xenobiotic stimulus
          response to virus
          response to herbicide
          positive regulation of gene expression
          siderophore transport
          positive regulation of cell projection organization
          innate immune response
          iron ion homeostasis
          protein homotrimerization
          cellular response to hydrogen peroxide
          extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          transporter activity
          iron ion binding
          protein binding
          small molecule binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115153 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115155 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115154 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115152 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115156 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070869 BC020275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070869 BC132069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070869 BC132071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070869 X14607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070869 AK149774.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10429 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102598 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070869 BC020275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070869 BC132069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070869 BC132071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070869 X14607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070869 AK149774.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P510156 BC020275.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P510156 BC132069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P510156 BC132071.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P510156 S82469.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P510156 X14607.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21115124 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102598 - -
          Whole Mouse Genome G4122A A_51_P510156 BC020275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P510156 BC132069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P510156 BC132071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P510156 S82469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P510156 X14607.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129241 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102598 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01478840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02041218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01478843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01478844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02041213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01430775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02041230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01478836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01478858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02041217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115153 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115154 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115152 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21115155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21115083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01430773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01356998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01356997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01698015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01430769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01430776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21115119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481628 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481633 - -
          Affymetrix HT MG-430 PM Array Plate 1427747_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481627 - -
          Mouse Genome 430 2.0 Array 1427747_PM_a_at - -
          Mouse Genome 430 2.0 Array 1427747_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16819_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16819_at - -
          Affymetrix Mouse Clariom S Array PSR0200028082.mm.1 TC0200003303.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028082.mm.1 NM_008491.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028082.mm.1 NM_008491.1 Refseq
          Mouse Expression Set 430 1427747_PM_a_at - -
          Mouse Expression Set 430 1427747_a_at - -
          Murine Genome U74v2 Set 160564_at - -
          Murine Genome U74v2 Set 161684_PM_r_at - -
          Murine Genome U74v2 Set 160564_PM_at - -
          Murine Genome U74v2 Set 161684_r_at - -
          Mouse Genome 430A 2.0 Array 1427747_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427747_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427747_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427747_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2510112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712075 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2712075 - -
          mouse-6 v1.1 (Illumina) scl19446.4_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 2510112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19446.4_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2712075 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712075 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19446.4_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0