Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00620423_m1
          See other HMCES GE Assays ›
          Gene Symbol:
          Hmces
          Gene Name:
          5-hydroxymethylcytosine (hmC) binding, ES cell specific
          Gene Aliases:
          8430410A17Rik, C85376
          Chromosome Location:
          Chr.6: 87913907 - 87936614 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00620423_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmces
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173737.2 NP_776098.1 6 - 7 886 58
          GenBank mRNA AK010550.1 - 4 - 5 652 58
          AK018400.1 - 5 - 6 1034 58
          AK215958.1 - 1 - 2 41 58
          BC024401.1 - 6 - 7 886 5026919 58
          BC064070.1 - 6 - 7 869 3371288 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001020.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmces

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          molecular_function
          DNA binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618852 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618851 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618847 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100963 BC064070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100963 AK010550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100963 AK018400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100963 NM_173737.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100963 BC024401.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44234 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160765 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118497 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100963 BC064070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100963 AK010550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100963 AK018400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100963 NM_173737.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100963 BC024401.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24618852 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118497 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05972748 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160765 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05972784 - -
          Whole Mouse Genome G4122A A_52_P504400 BC064070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504400 AK010550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504400 AK018400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504400 NM_173737.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504400 AK215958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298323 BC064070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298323 AK010550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298323 AK018400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298323 NM_173737.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298323 AK215958.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138545 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118497 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160765 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05128848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05545519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05545515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05128843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05128841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618851 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618852 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24618847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24618849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05415590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05128847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05539215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05844779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05116228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05128843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05539209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05790135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24618875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539851 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539853 - -
          Affymetrix HT MG-430 PM Array Plate 1423786_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539850 - -
          Mouse Genome 430 2.0 Array 1423786_at - -
          Mouse Genome 430 2.0 Array 1423786_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232210_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232210_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600008017.mm.1 TC0600001020.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008017.mm.1 NM_173737.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008017.mm.1 NM_173737.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008018.mm.1 TC0600001020.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008018.mm.1 NM_173737.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008018.mm.1 NM_173737.2 Refseq
          Mouse Expression Set 430 1423786_PM_at - -
          Mouse Expression Set 430 1423786_at - -
          Murine Genome U74v2 Set 160684_PM_at - -
          Murine Genome U74v2 Set 160684_at - -
          Murine Genome U74v2 Set 168394_PM_f_at - -
          Murine Genome U74v2 Set 168394_f_at - -
          Mouse Genome 430A 2.0 Array 1423786_at - -
          Mouse Genome 430A 2.0 Array 1423786_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423786_at - -
          Affymetrix HT Mouse Genome 430A Array 1423786_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645588 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6590128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604264 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604264 - -
          mouse-6 v1.1 (Illumina) scl29779.8.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 6590128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29779.8.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2604263 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604263 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29779.8.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-ndzlw:80/100.66.129.171:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0