Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100020486.mm.1 |
TC1100002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020486.mm.1 |
NM_001164620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020486.mm.1 |
NM_001164620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020486.mm.1 |
NM_177616.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020486.mm.1 |
NM_177616.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020488.mm.1 |
TC1100002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020488.mm.1 |
NM_001164620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020488.mm.1 |
NM_001164620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020488.mm.1 |
NM_177616.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020488.mm.1 |
NM_177616.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020487.mm.1 |
TC1100002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020487.mm.1 |
NM_001164620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020487.mm.1 |
NM_001164620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020487.mm.1 |
NM_177616.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020487.mm.1 |
NM_177616.3 |
Refseq |