Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00616327_m1
          See other TPRKB GE Assays ›
          Gene Symbol:
          Tprkb
          Gene Name:
          Tp53rk binding protein
          Gene Aliases:
          0610033G21Rik, 1810034M08Rik, Cgi-121, DRWMS1, mDRWMS1
          Chromosome Location:
          Chr.6: 85915278 - 85930284 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00616327_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tprkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170488.1 NP_001163959.1 6 - 7 726 89
          NM_176842.3 NP_789812.1 5 - 6 618 89
          GenBank mRNA BC024858.1 - 5 - 6 580 5357332 89
          BC027162.1 - 6 - 7 707 4951567 89
          BM948212.1 - 3 - 4 355 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000974.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tprkb

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          biological_process
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964218 NM_001170488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964218 NM_176842.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964213 BM948212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964213 BC024858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964213 NM_001170488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964213 NM_176842.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BM935112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BM948212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BC024858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BC027162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 NM_001170488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BM935112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BM948212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BC024858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 BC027162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P399677 NM_001170488.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131972 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163855 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110444 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123223 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964213 BM948212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964213 BC024858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964213 NM_001170488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964213 NM_176842.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964218 NM_001170488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964218 NM_176842.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BM935112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BM948212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BC024858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BC027162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 NM_001170488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BM935112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BM948212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BC024858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 BC027162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P399677 NM_001170488.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24607145 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05967624 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163855 - -
          Whole Mouse Genome G4122A A_52_P399677 BM935112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BM948212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BC024858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BC027162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 NM_001170488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BM935112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BM948212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BC024858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 BC027162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399677 NM_001170488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400613 AK075592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400613 BC024858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400613 NM_001170488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400613 NM_176842.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P400613 AB052769.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140982 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05053905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05447687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05447690 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05447696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05447688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05447689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24607196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05161507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05250663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05813172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05858939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05858933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24607153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539658 - -
          Affymetrix HT MG-430 PM Array Plate 1424992_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425682_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1425410_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539653 - -
          Mouse Genome 430 2.0 Array 1424992_PM_at - -
          Mouse Genome 430 2.0 Array 1425410_at - -
          Mouse Genome 430 2.0 Array 1425682_PM_a_at - -
          Mouse Genome 430 2.0 Array 1425410_PM_at - -
          Mouse Genome 430 2.0 Array 1425682_a_at - -
          Mouse Genome 430 2.0 Array 1424992_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69786_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69786_at - -
          Affymetrix Mouse Clariom S Array PSR0600007535.mm.1 TC0600000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007535.mm.1 NM_001170488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007535.mm.1 NM_001170488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007535.mm.1 NM_176842.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007535.mm.1 NM_176842.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007531.mm.1 TC0600000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007531.mm.1 NM_001170488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007531.mm.1 NM_001170488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007531.mm.1 NM_176842.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007531.mm.1 NM_176842.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007536.mm.1 TC0600000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007536.mm.1 NM_001170488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007536.mm.1 NM_001170488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007536.mm.1 NM_176842.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007536.mm.1 NM_176842.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007537.mm.1 TC0600000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007537.mm.1 NM_001170488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007537.mm.1 NM_001170488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007537.mm.1 NM_176842.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007537.mm.1 NM_176842.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007534.mm.1 TC0600000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007534.mm.1 NM_001170488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007534.mm.1 NM_001170488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007534.mm.1 NM_176842.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007534.mm.1 NM_176842.3 Refseq
          Mouse Expression Set 430 1425682_PM_a_at - -
          Mouse Expression Set 430 1424992_at - -
          Mouse Expression Set 430 1425682_a_at - -
          Mouse Expression Set 430 1425410_PM_at - -
          Mouse Expression Set 430 1425410_at - -
          Mouse Expression Set 430 1424992_PM_at - -
          Mouse Genome 430A 2.0 Array 1425410_PM_at - -
          Mouse Genome 430A 2.0 Array 1425410_at - -
          Mouse Genome 430A 2.0 Array 1424992_at - -
          Mouse Genome 430A 2.0 Array 1425682_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424992_PM_at - -
          Mouse Genome 430A 2.0 Array 1425682_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424992_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425682_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425410_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425682_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425410_at - -
          Affymetrix HT Mouse Genome 430A Array 1424992_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460601 - -
          MouseWG-6 v2 Expression BeadChip 101190086 - -
          MouseWG-6 v2 Expression BeadChip 3360193 - -
          Illumina mouse-6 v1.0 expression beadchip 460601 - -
          Illumina mouse-6 v1.0 expression beadchip 101190086 - -
          Illumina mouse-6 v1.0 expression beadchip 3360193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738991 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644950 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249173 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644950 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738991 - -
          mouse-6 v1.1 (Illumina) scl0069786.1_6-S - -
          mouse-6 v1.1 (Illumina) scl069786.2_30-S - -
          mouse-6 v1.1 (Illumina) scl0001238.1_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360193 - -
          Illumina Sentrix mouse-6.v1 bead array 101190086 - -
          Illumina Sentrix mouse-6.v1 bead array 460601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069786.2_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069786.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460601 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101190086 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2811240 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2644950 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2738991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2644950 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2811240 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0069786.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0