Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553577_m1
          See other COQ6 GE Assays ›
          Gene Symbol:
          Coq6
          Gene Name:
          coenzyme Q6 homolog (yeast)
          Gene Aliases:
          5930427M12Rik, AW742344
          Chromosome Location:
          Chr.12: 84361756 - 84373796 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553577_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coq6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172582.3 NP_766170.2 11 - 12 1485 85
          GenBank mRNA AK077863.1 - 11 - 12 1409 85
          AK131889.1 - 11 - 12 1447 85
          AK167380.1 - 11 - 12 1413 85
          AK170311.1 - 10 - 11 1950 85
          AK191776.1 - 2 - 3 255 85
          BC024135.1 - 11 - 12 1386 85
          BC051055.1 - 11 - 12 1401 85
          BC057914.1 - 8 - 9 1149 85
          BC088736.1 - 7 - 8 2734 85
          BC106183.1 - 11 - 12 1469 85
          BC132168.1 - 11 - 12 1485 40130445 85
          BC132170.1 - 11 - 12 1485 40130447 85

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coq6

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquinone biosynthetic process
          metabolic process
          secondary metabolite biosynthetic process
          molecular_function
          monooxygenase activity
          3-(3-hydroxyphenyl)propionate hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          4-chlorobenzaldehyde oxidase activity
          3,5-xylenol methylhydroxylase activity
          phenylacetate hydroxylase activity
          4-nitrophenol 4-monooxygenase activity
          dimethyl sulfide monooxygenase activity
          alpha-pinene monooxygenase [NADH] activity
          phenanthrene 9,10-monooxygenase activity
          1-hydroxy-2-naphthoate hydroxylase activity
          toluene 4-monooxygenase activity
          xylene monooxygenase activity
          dibenzothiophene monooxygenase activity
          6-hydroxy-3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
          chlorophenol 4-monooxygenase activity
          carbon disulfide oxygenase activity
          toluene 2-monooxygenase activity
          1-hydroxy-2-oxolimonene 1,2-monooxygenase activity
          phenanthrene 1,2-monooxygenase activity
          tetrahydrofuran hydroxylase activity
          styrene monooxygenase activity
          toluene-4-sulfonate monooxygenase activity
          toluene-sulfonate methyl-monooxygenase activity
          3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
          2-hydroxy-phenylacetate hydroxylase activity
          2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA 1,2-monooxygenase activity
          phenanthrene 3,4-monooxygenase activity
          toluene 3-monooxygenase activity
          4-hydroxyphenylacetate,NADH:oxygen oxidoreductase (3-hydroxylating) activity
          limonene monooxygenase activity
          2-methylnaphthalene hydroxylase activity
          1-methylnaphthalene hydroxylase activity
          bisphenol A hydroxylase A activity
          salicylate 5-hydroxylase activity
          isobutylamine N-hydroxylase activity
          branched-chain dodecylbenzene sulfonate monooxygenase activity
          3-HSA hydroxylase activity
          4-hydroxypyridine-3-hydroxylase activity
          2-octaprenyl-3-methyl-6-methoxy-1,4-benzoquinol hydroxylase activity
          6-hydroxynicotinate 3-monooxygenase activity
          flavin adenine dinucleotide binding
          tocotrienol omega-hydroxylase activity
          thalianol hydroxylase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056419 BC088736.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056419 BC106183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056419 BC051055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056419 BC057914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056419 BC024135.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64703 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142136 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124313 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056419 BC088736.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056419 BC106183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056419 BC051055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056419 BC057914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056419 BC024135.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116479 BC132170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116479 BC088736.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116479 BC106183.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116479 BC132168.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116479 BC051055.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28597841 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07690363 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124313 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142136 - -
          Whole Mouse Genome G4122A A_51_P116479 BC132170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P116479 BC088736.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P116479 BC106183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P116479 BC132168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P116479 BC051055.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135757 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142136 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09776112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28597885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10204665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09776100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10125352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10420041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10267918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10125351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10126716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10285971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10267907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10285967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10308607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28597855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10397217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397222 - -
          Affymetrix HT MG-430 PM Array Plate 1426902_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10397216 - -
          Mouse Genome 430 2.0 Array 1426902_at - -
          Mouse Genome 430 2.0 Array 1426902_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217707_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217707_PM_at - -
          Mouse Expression Set 430 1426902_PM_at - -
          Mouse Expression Set 430 1426902_at - -
          Murine Genome U74v2 Set 170810_PM_at - -
          Murine Genome U74v2 Set 170810_at - -
          Murine Genome U74v2 Set 169152_PM_r_at - -
          Murine Genome U74v2 Set 169152_r_at - -
          Mouse Genome 430A 2.0 Array 1426902_at - -
          Mouse Genome 430A 2.0 Array 1426902_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426902_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426902_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2760162 - -
          Illumina mouse-6 v1.0 expression beadchip 5670014 - -
          Illumina mouse-6 v1.0 expression beadchip 2760162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670014 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766720 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229589 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229589 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2766720 - -
          mouse-6 v1.1 (Illumina) scl0002298.1_37-S - -
          mouse-6 v1.1 (Illumina) scl42976.13.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670014 - -
          Illumina Sentrix mouse-6.v1 bead array 2760162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002298.1_37-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42976.13.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670014 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2766720 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229589 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2898403 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766720 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229589 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2898403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl42976.13.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7dfb8678b5-vqhv7:80/100.66.129.46:80.
          git-commit: 632cddff25488e9c1007a93c87c1899f725eef0d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0