Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530295_s1
          See other OLFR613 GE Assays ›
          Gene Symbol:
          Olfr613
          Gene Name:
          olfactory receptor 613
          Gene Aliases:
          MOR20-1, Olfr613-ps1, Olfr614
          Chromosome Location:
          Chr.7: 103550368 - 103555504 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530295_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr613
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147100.3 NP_667311.3 1 2172 73
          GenBank mRNA AK156691.1 - 1 2171 73
          BC145889.1 - 1 784 40131192 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001497.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr613

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231113 NM_147100.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231113 AK156691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231113 BC145889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231113 NM_147100.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231113 AK156691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231113 BC145889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339054 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119502 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021108 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231113 NM_147100.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231113 AK156691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231113 BC145889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231113 NM_147100.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231113 AK156691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231113 BC145889.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231113 NM_147100.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231113 AK156691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231113 BC145889.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231113 NM_147100.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231113 AK156691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231113 BC145889.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119502 - -
          Whole Mouse Genome G4122A A_51_P231113 NM_147100.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231113 AK156691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231113 BC145889.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231113 NM_147100.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231113 AK156691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231113 BC145889.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138228 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119502 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06404424 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06404423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06634638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06634639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06634635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06024106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06074173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06074175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06634634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555798 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555795 - -
          Affymetrix Mouse Clariom S Array PSR0700012758.mm.1 TC0700001497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012758.mm.1 NM_147100.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012758.mm.1 NM_147100.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012757.mm.1 TC0700001497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012757.mm.1 NM_147100.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012757.mm.1 NM_147100.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 365190 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681275 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2681275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0