Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167884.1 | NP_001161356.1 | 10 - 11 | 1608 | 100 | |
NM_001167885.1 | NP_001161357.1 | 7 - 8 | 1306 | 100 | ||
NM_001167886.1 | NP_001161358.1 | 5 - 6 | 1177 | 100 | ||
NM_001167887.1 | NP_001161359.1 | 6 - 7 | 1246 | 100 | ||
NM_001167888.1 | NP_001161360.1 | 7 - 8 | 1306 | 100 | ||
NM_001167889.1 | NP_001161361.1 | 6 - 7 | 1246 | 100 | ||
NM_144871.4 | NP_659120.3 | 6 - 7 | 1237 | 100 | ||
GenBank mRNA | AK082660.1 | - | 5 - 6 | 926 | 100 | |
AK083227.1 | - | 6 - 7 | 1238 | 100 | ||
AK087267.1 | - | 10 - 11 | 1510 | 100 | ||
AK139255.1 | - | 5 - 6 | 811 | 100 | ||
AK152179.1 | - | 6 - 7 | 850 | 100 | ||
AY555192.1 | - | 5 - 6 | 936 | 100 | ||
BC011214.1 | - | 6 - 7 | 872 | 100 | ||
BC075709.1 | - | 4 - 5 | 723 | 3661032 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |