Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167884.1 | NP_001161356.1 | 9 - 10 | 1447 | 86 | |
NM_001167885.1 | NP_001161357.1 | 6 - 7 | 1145 | 86 | ||
NM_001167887.1 | NP_001161359.1 | 5 - 6 | 1085 | 86 | ||
NM_001167888.1 | NP_001161360.1 | 6 - 7 | 1145 | 86 | ||
NM_001167889.1 | NP_001161361.1 | 5 - 6 | 1085 | 86 | ||
GenBank mRNA | AK054093.1 | - | 4 - 5 | 681 | 86 | |
AK082660.1 | - | 4 - 5 | 765 | 86 | ||
AK087267.1 | - | 9 - 10 | 1349 | 86 | ||
AK139255.1 | - | 4 - 5 | 650 | 86 | ||
AK152179.1 | - | 5 - 6 | 689 | 86 | ||
AY555192.1 | - | 4 - 5 | 775 | 86 | ||
BC011214.1 | - | 5 - 6 | 711 | 86 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |