Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163332.1 | NP_001156804.1 | 3 - 4 | 280 | 147 | |
NM_001163333.1 | NP_001156805.1 | 3 - 4 | 272 | 147 | ||
NM_030249.4 | NP_084525.1 | 3 - 4 | 388 | 147 | ||
GenBank mRNA | AK029901.1 | - | 3 - 4 | 389 | 147 | |
AK030438.1 | - | 3 - 4 | 253 | 147 | ||
AK051259.1 | - | 3 - 4 | 252 | 147 | ||
AK129359.1 | - | 3 - 4 | 278 | 147 | ||
AK145085.1 | - | 3 - 4 | 251 | 147 | ||
BC003236.1 | - | 3 - 4 | 242 | 3498078 | 147 | |
CV562515.1 | - | 3 - 4 | 272 | 147 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |