Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517808_m1
          See other NAT8 GE Assays ›
          Gene Symbol:
          Nat8
          Gene Name:
          N-acetyltransferase 8 (GCN5-related, putative)
          Gene Aliases:
          0610037O16Rik, CCNAT, Cml4
          Chromosome Location:
          Chr.6: 85830387 - 85836636 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517808_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nat8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023455.3 NP_075944.1 1 - 2 103 74
          GenBank mRNA AF163317.1 - 1 - 2 74
          AK002784.1 - 1 - 2 45 74
          BC019517.1 - 1 - 2 295 4236543 74
          BY081766.1 - 1 - 2 84 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF163317.1, AK002784.1, BC019517.1, NM_023455.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nat8

          Gene Ontology Categories:

          Function(s) Process(es)

          gastrulation with mouth forming second
          glutathione metabolic process
          negative regulation of cell adhesion
          positive regulation of gene expression
          peptidyl-lysine N6-acetylation
          negative regulation of apoptotic process
          amyloid-beta metabolic process
          dihydrolipoamide branched chain acyltransferase activity
          lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          sterol O-acyltransferase activity
          N-acetyltransferase activity
          O-acyltransferase activity
          palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
          carnitine O-acyltransferase activity
          acetyltransferase activity
          C-acyltransferase activity
          palmitoyltransferase activity
          N-acyltransferase activity
          acylglycerol O-acyltransferase activity
          serine O-acyltransferase activity
          O-acetyltransferase activity
          O-octanoyltransferase activity
          octanoyltransferase activity
          O-palmitoyltransferase activity
          S-acyltransferase activity
          S-acetyltransferase activity
          S-malonyltransferase activity
          malonyltransferase activity
          C-palmitoyltransferase activity
          transferase activity
          acyltransferase activity
          succinyltransferase activity
          N-succinyltransferase activity
          O-succinyltransferase activity
          S-succinyltransferase activity
          sinapoyltransferase activity
          O-sinapoyltransferase activity
          peptidyl-lysine N6-myristoyltransferase activity
          peptidyl-lysine N6-palmitoyltransferase activity
          benzoyl acetate-CoA thiolase activity
          3-hydroxybutyryl-CoA thiolase activity
          3-ketopimelyl-CoA thiolase activity
          N-palmitoyltransferase activity
          myristoyltransferase activity
          acyl-CoA N-acyltransferase activity
          protein-cysteine S-myristoyltransferase activity
          protein-cysteine S-acyltransferase activity
          dihydrolipoamide S-acyltransferase activity
          glucosaminyl-phosphatidylinositol O-acyltransferase activity
          ergosterol O-acyltransferase activity
          lanosterol O-acyltransferase activity
          naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
          2-methylhexanoyl-CoA C-acetyltransferase activity
          butyryl-CoA 2-C-propionyltransferase activity
          2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
          alpha-aminoadipate acetyltransferase activity
          keto acid formate lyase activity
          Ras palmitoyltransferase activity
          azetidine-2-carboxylic acid acetyltransferase activity
          cysteine-S-conjugate N-acetyltransferase activity
          peptidyl-lysine acetyltransferase activity
          acetyl-CoA:L-lysine N6-acetyltransferase
          UDP-3-O-(R-3-hydroxymyristoyl)-glucosamine N-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 NM_023455.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 AF163317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 AK002784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 BC019517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 NM_023455.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 AF163317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 AK002784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479230 BC019517.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300110 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140290 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 NM_023455.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 AF163317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 AK002784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 BC019517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 NM_023455.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 AF163317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 AK002784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479230 BC019517.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 NM_023455.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 AF163317.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 AK002784.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 BC019517.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 NM_023455.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 AF163317.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 AK002784.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479230 BC019517.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24606751 - -
          Whole Mouse Genome G4122A A_51_P479230 NM_023455.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 AF163317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 AK002784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 BC019517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 NM_023455.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 AF163317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 AK002784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479230 BC019517.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05650886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05650866 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05650887 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05650879 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05650882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05650909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05541174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05541179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05650864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606780 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545879 - -
          Affymetrix HT MG-430 PM Array Plate 1419520_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545877 - -
          Mouse Genome 430 2.0 Array 1419520_PM_at - -
          Mouse Genome 430 2.0 Array 1419520_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68396_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68396_at - -
          Mouse Expression Set 430 1419520_PM_at - -
          Mouse Expression Set 430 1419520_at - -
          Murine Genome U74v2 Set 167026_at - -
          Murine Genome U74v2 Set 167026_PM_at - -
          Mouse Genome 430A 2.0 Array 1419520_PM_at - -
          Mouse Genome 430A 2.0 Array 1419520_at - -
          Affymetrix HT Mouse Genome 430A Array 1419520_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419520_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 821564 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7050056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216539 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216539 - -
          mouse-6 v1.1 (Illumina) scl068396.1_227-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068396.1_227-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216539 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216539 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0