Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018883.2 | NP_061371.2 | 5 - 6 | 681 | 123 | |
GenBank mRNA | AF117384.1 | - | 5 - 6 | 636 | 123 | |
AK144483.1 | - | 5 - 6 | 682 | 123 | ||
BC017529.1 | - | 5 - 6 | 646 | 4924656 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |