Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513834_m1
          See other 2210407C18RIK GE Assays ›
          Gene Symbol:
          2210407C18Rik
          Gene Name:
          RIKEN cDNA 2210407C18 gene
          Gene Aliases:
          Ep1
          Chromosome Location:
          Chr.11: 58608204 - 58613492 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513834_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2210407C18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC019553.1 - 5 - 6 550 4456658 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002829.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992546 BC019553.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22368 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147313 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992546 BC019553.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831868 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147313 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07357759 - -
          Whole Mouse Genome G4122A A_51_P316673 BC019553.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119955 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09731482 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09731489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09731483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09053406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09266553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09698182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08981972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09469819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09184583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09052796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09469818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09050938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09009647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09731487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09450710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08981968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831838 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386201 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386198 - -
          Affymetrix HT MG-430 PM Array Plate 1425233_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10386197 - -
          Mouse Genome 430 2.0 Array 1425233_at - -
          Mouse Genome 430 2.0 Array 1425233_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78354_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78354_at - -
          Affymetrix Mouse Clariom S Array PSR1100025568.mm.1 TC1100002829.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025568.mm.1 NM_144544.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025568.mm.1 NM_144544.2 Refseq
          Mouse Expression Set 430 1425233_PM_at - -
          Mouse Expression Set 430 1425233_at - -
          Mouse Genome 430A 2.0 Array 1425233_PM_at - -
          Mouse Genome 430A 2.0 Array 1425233_at - -
          Affymetrix HT Mouse Genome 430A Array 1425233_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425233_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633579 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226767 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226767 - -
          mouse-6 v1.1 (Illumina) scl40181.5.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40181.5.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226767 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226767 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40181.5.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0