Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512131_g1
          See other RAB10OS GE Assays ›
          Gene Symbol:
          Rab10os
          Gene Name:
          RAB10, member RAS oncogene family, opposite strand
          Gene Aliases:
          1700012B15Rik, 1810036A22Rik
          Chromosome Location:
          Chr.12: 3235791 - 3250374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512131_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab10os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015551.1 2 - 3 268 106
          GenBank mRNA AK005738.1 - 2 - 3 221 106
          AK005891.1 - 2 - 3 548 106
          AK007702.1 - 2 - 3 269 106
          AK171768.1 - 2 - 3 548 106
          BC038315.1 - 2 - 3 219 106
          BC060685.1 - 2 - 3 1802 106
          BC062897.1 - 2 - 3 1803 106
          BC080728.1 - 1 - 2 1889 106
          BC099424.1 - 2 - 3 531 106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF017478.1 - 2 - 3 Chr. 12 - 3237320 - 3238274 [-] on Build GRCm38 106
          GenBankmRNA BY564105.1 - 1 - 2 105 Chr. 12 - 3237309 - 3238282 [+] on Build GRCm38 106
          GenBankmRNA CK792083.1 - 2 - 3 542 Chr. 12 - 3235530 - 3238113 [+] on Build GRCm38 106
          GenBankmRNA BG921953.1 - 1 - 2 144 Chr. 12 - 3237280 - 3249944 [+] on Build GRCm38 106
          GenBankmRNA BY707724.1 - 2 - 3 266 Chr. 12 - 3235792 - 3250282 [+] on Build GRCm38 106
          GenBankmRNA BY558274.1 - 1 - 2 132 Chr. 12 - 3237285 - 3238284 [+] on Build GRCm38 107
          GenBankmRNA BY521541.1 - 1 - 2 108 Chr. 12 - 3237307 - 3238273 [+] on Build GRCm38 106
          GenBankmRNA BY705904.1 - 2 - 3 221 Chr. 12 - 3236611 - 3249920 [+] on Build GRCm38 106
          GenBankmRNA BI082869.1 - 234 Chr. - - [-] on Build 107
          GenBankmRNA CV559944.1 - 2 - 3 533 Chr. 12 - 3235526 - 3238180 [+] on Build GRCm38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Downstream RAB10,
          member RAS oncogene family
          Rab10 AW107754 9.8kb Anti-sense Chr. 12 - 3247428 - 3309969 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000005.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0600000319.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200000027.mm.1 TC1200000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000027.mm.1 NR_015551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000027.mm.1 NR_015551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000029.mm.1 TC1200000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000029.mm.1 NR_015551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000029.mm.1 NR_015551.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-57bdf9945f-4p2c8:80/100.66.134.79:80.
          git-commit: baa7f4fdc4fe37bd11ed6fe4edff1157ae59fd06
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0