Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028722.2 | NP_082998.2 | 2 - 3 | 1064 | 95 | |
GenBank mRNA | AK014463.1 | - | 2 - 3 | 1317 | 95 | |
AK015496.1 | - | 2 - 3 | 328 | 95 | ||
AK053719.1 | - | 2 - 3 | 1034 | 95 | ||
AK161412.1 | - | 2 - 3 | 307 | 95 | ||
BC043039.1 | - | 2 - 3 | 1042 | 5701204 | 95 | |
BC055715.1 | - | 2 - 3 | 1016 | 6414174 | 95 | |
BC058629.1 | - | 2 - 3 | 1064 | 6402215 | 95 | |
DQ387855.1 | - | 2 - 3 | 327 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |