Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505035_g1
          See other FBXL15 GE Assays ›
          Gene Symbol:
          Fbxl15
          Gene Name:
          F-box and leucine-rich repeat protein 15
          Gene Aliases:
          0710008C12Rik, Fbxo37
          Chromosome Location:
          Chr.19: 46328059 - 46330446 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505035_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxl15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133694.2 NP_598455.2 1 - 2 341 92
          GenBank mRNA AK003032.1 - 1 - 2 92
          AK145009.1 - 1 - 2 299 92
          AK172339.1 - 2 - 3 238 92
          BC016499.1 - 1 - 2 316 4221584 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000691.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK172339.1, BC016499.1, NM_133694.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxl15

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          protein ubiquitination
          bone mineralization
          positive regulation of BMP signaling pathway
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          ubiquitin-protein transferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900006180.mm.1 TC1900000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900006180.mm.1 NM_133694.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006180.mm.1 NM_133694.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006181.mm.1 TC1900000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900006181.mm.1 NM_133694.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006181.mm.1 NM_133694.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006182.mm.1 TC1900000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900006182.mm.1 NM_133694.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006182.mm.1 NM_133694.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006183.mm.1 TC1900000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900006183.mm.1 NM_133694.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006183.mm.1 NM_133694.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-57bdf9945f-4p2c8:80/100.66.134.79:80.
          git-commit: baa7f4fdc4fe37bd11ed6fe4edff1157ae59fd06
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0