Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016638.mm.1 |
TC1500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016638.mm.1 |
NM_026817.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016638.mm.1 |
NM_026817.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016635.mm.1 |
TC1500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016635.mm.1 |
NM_026817.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016635.mm.1 |
NM_026817.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016636.mm.1 |
TC1500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016636.mm.1 |
NM_026817.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016636.mm.1 |
NM_026817.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016634.mm.1 |
TC1500002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016634.mm.1 |
NM_026817.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016634.mm.1 |
NM_026817.3 |
Refseq |