Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503454_g1
          See other MLH1 GE Assays ›
          Gene Symbol:
          Mlh1
          Gene Name:
          mutL homolog 1 (E. coli)
          Gene Aliases:
          1110035C23Rik, AI317206, AI325952, AI561766
          Chromosome Location:
          Chr.9: 111228228 - 111271786 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503454_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mlh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026810.2 NP_081086.2 17 - 18 2083 112
          GenBank mRNA AF250844.1 - 17 - 18 1999 112
          AK004105.1 - 8 - 9 1173 112
          AK145646.1 - 17 - 18 2030 112
          AK163861.1 - 16 - 17 3249 112
          AK168849.1 - 17 - 18 2022 112
          BC021815.1 - 17 - 18 2037 4020489 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003128.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mlh1

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA poly(A) tail shortening
          resolution of meiotic recombination intermediates
          somatic recombination of immunoglobulin genes involved in immune response
          DNA repair
          mismatch repair
          double-strand break repair via nonhomologous end joining
          cellular response to DNA damage stimulus
          cell cycle
          male meiosis chromosome segregation
          homologous chromosome pairing at meiosis
          reciprocal meiotic recombination
          male meiotic nuclear division
          spermatogenesis
          intrinsic apoptotic signaling pathway in response to DNA damage
          female meiosis chromosome segregation
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          meiotic metaphase I plate congression
          meiotic chromosome segregation
          homologous chromosome segregation
          isotype switching
          negative regulation of mitotic recombination
          oogenesis
          meiotic spindle midzone assembly
          meiotic cell cycle
          DNA binding
          single-stranded DNA binding
          ATP binding
          ATP hydrolysis activity
          mismatched DNA binding
          guanine/thymine mispair binding
          MutSalpha complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900026480.mm.1 TC0900003128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026480.mm.1 NM_026810.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026480.mm.1 NM_026810.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026482.mm.1 TC0900003128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026482.mm.1 NM_026810.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026482.mm.1 NM_026810.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026483.mm.1 TC0900003128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026483.mm.1 NM_026810.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026483.mm.1 NM_026810.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0