Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104616.1 | NP_001098086.1 | 3 - 4 | 249 | 112 | |
NM_001104617.1 | NP_001098087.1 | 3 - 4 | 655 | 112 | ||
NM_009041.3 | NP_033067.2 | 3 - 4 | 655 | 112 | ||
GenBank mRNA | AK016711.1 | - | 3 - 4 | 326 | 112 | |
AK088292.1 | - | 3 - 4 | 362 | 112 | ||
AK146423.1 | - | 3 - 4 | 313 | 112 | ||
AK156370.1 | - | 3 - 4 | 341 | 112 | ||
AK162204.1 | - | 3 - 4 | 227 | 112 | ||
AK168449.1 | - | 3 - 4 | 326 | 112 | ||
BC002041.1 | - | 3 - 4 | 300 | 112 | ||
BC026786.1 | - | 3 - 4 | 328 | 112 | ||
BC034438.1 | - | 3 - 4 | 481 | 112 | ||
BC053417.1 | - | 3 - 4 | 329 | 6707873 | 112 | |
DV662898.1 | - | 3 - 4 | 249 | 112 | ||
X60672.1 | - | 3 - 4 | 268 | 112 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |