Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499576_m1
          See other RAB14 GE Assays ›
          Gene Symbol:
          Rab14
          Gene Name:
          RAB14, member RAS oncogene family
          Gene Aliases:
          0610030G24Rik, 2810475J17Rik, A830021G03Rik, AI314285, AI649155, D030017L14Rik
          Chromosome Location:
          Chr.2: 35180205 - 35201431 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499576_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026697.3 NP_080973.1 1 - 2 362 75
          GenBank mRNA AJ968955.1 - 1 - 2 217 75
          AJ968956.1 - 1 - 2 217 75
          AJ968957.1 - 1 - 2 217 75
          AJ968958.1 - 1 - 2 217 75
          AJ968959.1 - 1 - 2 217 75
          AJ968960.1 - 1 - 2 217 75
          AK002704.1 - 1 - 2 234 75
          AK013410.1 - 1 - 2 215 75
          AK133963.1 - 1 - 2 262 75
          AK135751.1 - 1 - 2 251 75
          AK160859.1 - 1 - 2 362 75
          AK165812.1 - 1 - 2 209 75
          AK168551.1 - 1 - 2 219 75
          AK168867.1 - 1 - 2 231 75
          AK169396.1 - 1 - 2 233 75
          BC025139.1 - 1 - 2 233 3980228 75
          BC056648.1 - 1 - 2 235 5034519 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003360.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ968955.1, AJ968956.1, AJ968957.1, AJ968958.1, AJ968959.1, AJ968960.1, AK002704.1, AK133963.1, AK160859.1, AK165812.1, AK168551.1, AK168867.1, AK169396.1, BC025139.1, BC056648.1, NM_026697.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab14

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          Golgi to endosome transport
          small GTPase mediated signal transduction
          body fluid secretion
          metabolic process
          fibroblast growth factor receptor signaling pathway
          embryo development
          protein transport
          endocytic recycling
          Rab protein signal transduction
          regulation of protein localization
          apical protein localization
          phagolysosome assembly involved in apoptotic cell clearance
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          myosin V binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200028915.mm.1 TC0200003360.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028915.mm.1 NM_026697.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028915.mm.1 NM_026697.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028922.mm.1 TC0200003360.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028922.mm.1 NM_026697.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028922.mm.1 NM_026697.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028924.mm.1 TC0200003360.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028924.mm.1 NM_026697.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028924.mm.1 NM_026697.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028928.mm.1 TC0200003360.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028928.mm.1 NM_026697.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028928.mm.1 NM_026697.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 675318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0