Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019097.mm.1 |
TC0200002381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019097.mm.1 |
NM_025516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019097.mm.1 |
NM_025516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019095.mm.1 |
TC0200002381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019095.mm.1 |
NM_025516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019095.mm.1 |
NM_025516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019105.mm.1 |
TC0200002381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019105.mm.1 |
NM_025516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019105.mm.1 |
NM_025516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019099.mm.1 |
TC0200002381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019099.mm.1 |
NM_025516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019099.mm.1 |
NM_025516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019104.mm.1 |
TC0200002381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019104.mm.1 |
NM_025516.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019104.mm.1 |
NM_025516.4 |
Refseq |