Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290805.1 | NP_001277734.1 | 10 - 11 | 1534 | 67 | |
NM_001290806.1 | NP_001277735.1 | 11 - 12 | 1522 | 67 | ||
NM_008443.4 | NP_032469.2 | 11 - 12 | 1543 | 67 | ||
GenBank mRNA | AK147078.1 | - | 10 - 11 | 1484 | 67 | |
AK169425.1 | - | 11 - 12 | 1496 | 67 | ||
BC052707.1 | - | 11 - 12 | 1509 | 5702415 | 67 | |
D12645.1 | - | 11 - 12 | 1519 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |