Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489480_g1
          See other RAB10 GE Assays ›
          Gene Symbol:
          Rab10
          Gene Name:
          RAB10, member RAS oncogene family
          Gene Aliases:
          AW107754
          Chromosome Location:
          Chr.12: 3247428 - 3309969 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489480_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016676.5 NP_057885.1 4 - 5 970 109
          GenBank mRNA AB232604.1 - 4 - 5 419 109
          AF035646.1 - 4 - 5 660 109
          AK008725.1 - 4 - 5 960 109
          AK028320.1 - 4 - 5 941 109
          AK087964.1 - 4 - 5 932 109
          AK134988.1 - 4 - 5 890 109
          AK146357.1 - 4 - 5 971 109
          AK150319.1 - 4 - 5 930 109
          AK150384.1 - 4 - 5 624 109
          AK150791.1 - 4 - 5 920 109
          AK151473.1 - 4 - 5 624 109
          AK153315.1 - 4 - 5 930 109
          AK153325.1 - 4 - 5 920 109
          AK159383.1 - 4 - 5 935 109
          AK159845.1 - 4 - 5 935 109
          AK159988.1 - 4 - 5 929 109
          AK160960.1 - 4 - 5 925 109
          AK166010.1 - 4 - 5 944 109
          AK168320.1 - 4 - 5 930 109
          AK190935.1 - 3 - 4 272 109
          AK193085.1 - 3 - 4 253 109
          AK212462.1 - 2 - 3 204 109
          BC052735.1 - 920 109
          BC056374.1 - 4 - 5 921 6848840 109
          CT010394.1 - 4 - 5 419 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001339.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab10

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          Golgi to plasma membrane transport
          vesicle docking involved in exocytosis
          small GTPase mediated signal transduction
          axonogenesis
          metabolic process
          protein secretion
          protein transport
          vesicle-mediated transport
          endosomal transport
          regulation of exocytosis
          antigen processing and presentation
          Rab protein signal transduction
          cellular response to insulin stimulus
          establishment of neuroblast polarity
          endoplasmic reticulum tubular network organization
          protein localization to plasma membrane
          establishment of protein localization to membrane
          establishment of protein localization to endoplasmic reticulum membrane
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          myosin V binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200009609.mm.1 TC1200001339.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009609.mm.1 NM_016676.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009609.mm.1 NM_016676.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0