Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030469.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030469.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030469.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030472.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030472.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030472.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030471.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030471.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030471.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030467.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030467.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030467.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030463.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030463.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030463.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030465.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030465.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030465.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030468.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030468.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030468.mm.1 |
NM_008702.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030464.mm.1 |
TC1100003303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030464.mm.1 |
NM_008702.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030464.mm.1 |
NM_008702.3 |
Refseq |