Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400020336.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020336.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020336.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020336.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020336.mm.1 |
NM_053081.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020341.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020341.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020341.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020341.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020341.mm.1 |
NM_053081.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020335.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020335.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020335.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020335.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020335.mm.1 |
NM_053081.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020339.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020339.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020339.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020339.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020339.mm.1 |
NM_053081.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020343.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020343.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020343.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020343.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020343.mm.1 |
NM_053081.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020338.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020338.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020338.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020338.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020338.mm.1 |
NM_053081.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020344.mm.1 |
TC0400002514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020344.mm.1 |
NM_001163233.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020344.mm.1 |
NM_001163233.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020344.mm.1 |
NM_053081.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020344.mm.1 |
NM_053081.2 |
Refseq |