Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00467635_m1
          See other KLRB1F GE Assays ›
          Gene Symbol:
          Klrb1f
          Gene Name:
          killer cell lectin-like receptor subfamily B member 1F
          Gene Aliases:
          A630024B12Rik, Nkr-p1f, Nkrp1-f, Nkrp1f
          Chromosome Location:
          Chr.6: 129036902 - 129059724 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00467635_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klrb1f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153094.2 NP_694734.1 1 - 2 136 74
          NR_024262.1 1 - 2 136 74
          NR_024263.1 1 - 2 136 74
          GenBank mRNA AK041601.1 - 1 - 2 137 74
          AK154278.1 - 1 - 2 123 74
          AK170023.1 - 1 - 2 113 74
          AY029597.1 - 1 - 2 101 74
          BC107177.1 - 1 - 2 117 74
          BC107178.1 - 1 - 2 117 74
          BC127141.1 - 1 - 2 117 74
          BC127142.1 - 1 - 2 113 40050668 74
          DQ237931.1 - 1 - 2 89 74
          HQ713435.1 - 1 - 2 89 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC107177.1, BC107178.1, BC127141.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klrb1f

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding
          carbohydrate binding
          signaling receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 AK154278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 AK170023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 AK042352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 NM_153094.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 NR_024262.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 AK154278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 AK170023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 AK042352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 NM_153094.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625912 NR_024262.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41499 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145737 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119609 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 AK154278.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 AK170023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 AK042352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 NM_153094.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 NR_024262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 AK154278.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 AK170023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 AK042352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 NM_153094.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625912 NR_024262.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24841758 - -
          Whole Mouse Genome G4122A A_52_P680667 BC127142.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680667 DQ237931.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680667 BC127141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680667 AK154278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680667 AK170023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 AK154278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 AK170023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 AK042352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 NM_153094.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 NR_024262.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 AK154278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 AK170023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 AK042352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 NM_153094.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625912 NR_024262.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05122303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24841819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05693392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05007413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05775654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05607809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542147 - -
          Affymetrix HT MG-430 PM Array Plate 1425962_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457722_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10542140 - -
          Mouse Genome 430 2.0 Array 1425962_at - -
          Mouse Genome 430 2.0 Array 1457722_PM_at - -
          Mouse Genome 430 2.0 Array 1457722_at - -
          Mouse Genome 430 2.0 Array 1425962_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232408_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232408_at - -
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 TC0600001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 NM_153094.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 NM_153094.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 NR_024262.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 NR_024262.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 NR_024263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012438.mm.1 NR_024263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 TC0600001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 NM_153094.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 NM_153094.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 NR_024262.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 NR_024262.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 NR_024263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012437.mm.1 NR_024263.1 Refseq
          Mouse Expression Set 430 1425962_at - -
          Mouse Expression Set 430 1425962_PM_at - -
          Mouse Expression Set 430 1457722_PM_at - -
          Mouse Expression Set 430 1457722_at - -
          Mouse Genome 430A 2.0 Array 1425962_PM_at - -
          Mouse Genome 430A 2.0 Array 1425962_at - -
          Affymetrix HT Mouse Genome 430A Array 1425962_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425962_at - -
          Affymetrix HT Mouse Genome 430B Array 1457722_at - -
          Affymetrix HT Mouse Genome 430B Array 1457722_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496450 - -
          ABI Mouse Genome Survey Microarray 711738 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104230021 - -
          MouseWG-6 v2 Expression BeadChip 1690102 - -
          Illumina mouse-6 v1.0 expression beadchip 104230021 - -
          Illumina mouse-6 v1.0 expression beadchip 1690102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001690102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694646 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604651 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2499201 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694646 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604651 - -
          mouse-6 v1.1 (Illumina) scl29468.7_627-S - -
          mouse-6 v1.1 (Illumina) scl00232408.2_319-S - -
          Illumina Sentrix mouse-6.v1 bead array 104230021 - -
          Illumina Sentrix mouse-6.v1 bead array 1690102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00232408.2_319-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104230021 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2604651 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694646 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604651 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2499201 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0