Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462764_m1
          See other PXT1 GE Assays ›
          Gene Symbol:
          Pxt1
          Gene Name:
          peroxisomal, testis specific 1
          Gene Aliases:
          1700001G18Rik, Stepp
          Chromosome Location:
          Chr.17: 28933986 - 28942262 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462764_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pxt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153390.1 NP_700439.1 2 - 3 193 83
          GenBank mRNA AF486826.1 - 2 - 3 193 83
          AK005593.1 - 2 - 3 196 83
          BC100437.1 - 2 - 3 192 30915792 83

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF486826.1, BC100437.1, NM_153390.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pxt1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of apoptotic process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31125151 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31125150 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 AK005593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 AF486826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 BC100437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 NM_153390.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 AK005593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 AF486826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 BC100437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422194 NM_153390.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41874 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175292 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 AK005593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 AF486826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 BC100437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 NM_153390.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 AK005593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 AF486826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 BC100437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422194 NM_153390.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125103 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08752059 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175292 - -
          Whole Mouse Genome G4122A A_51_P422194 AK005593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 AF486826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 BC100437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 NM_153390.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 AK005593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 AF486826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 BC100437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422194 NM_153390.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126417 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175292 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13108418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13108417 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13108414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31125151 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31125150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13088649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13366434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13378150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13113460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13305859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13147453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13113454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125117 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449526 - -
          Affymetrix HT MG-430 PM Array Plate 1430015_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449523 - -
          Mouse Genome 430 2.0 Array 1430015_at - -
          Mouse Genome 430 2.0 Array 1430015_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69307_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69307_at - -
          Affymetrix Mouse Clariom S Array PSR1700016340.mm.1 TC1700001772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016340.mm.1 NM_153390.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016340.mm.1 NM_153390.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016341.mm.1 TC1700001772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016341.mm.1 NM_153390.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016341.mm.1 NM_153390.1 Refseq
          Mouse Expression Set 430 1430015_PM_at - -
          Mouse Expression Set 430 1430015_at - -
          Murine Genome U74v2 Set 168576_i_at - -
          Murine Genome U74v2 Set 168576_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1430015_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430015_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 516913 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2480592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2663291 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2663291 - -
          mouse-6 v1.1 (Illumina) scl50110.3.1_68-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50110.3.1_68-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2663291 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663291 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0