Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253783.1 | NP_001240712.1 | 6 - 7 | 581 | 61 | |
NM_001253784.1 | NP_001240713.1 | 5 - 6 | 559 | 61 | ||
NM_001281803.1 | NP_001268732.1 | 4 - 5 | 563 | 61 | ||
NM_054046.5 | NP_473387.1 | 4 - 5 | 472 | 61 | ||
GenBank mRNA | AK028726.1 | - | 5 - 6 | 531 | 61 | |
AK034028.1 | - | 438 | 61 | |||
AK046271.1 | - | 4 - 5 | 432 | 61 | ||
AK050405.1 | - | 5 - 6 | 530 | 61 | ||
AK081295.1 | - | 4 - 5 | 462 | 61 | ||
AK086174.1 | - | 4 - 5 | 433 | 61 | ||
AK087778.1 | - | 4 - 5 | 443 | 61 | ||
AK129019.1 | - | 4 - 5 | 514 | 61 | ||
AK133979.1 | - | 4 - 5 | 446 | 61 | ||
AK150334.1 | - | 4 - 5 | 408 | 61 | ||
AK150994.1 | - | 4 - 5 | 408 | 61 | ||
AK164147.1 | - | 4 - 5 | 444 | 61 | ||
BC003306.1 | - | 6 - 7 | 532 | 3154712 | 61 | |
BY713285.1 | - | 4 - 5 | 473 | 61 | ||
CX562217.1 | - | 4 - 5 | 525 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |