Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448076_s1
          See other OLFR54 GE Assays ›
          Gene Symbol:
          Olfr54
          Gene Name:
          olfactory receptor 54
          Gene Aliases:
          F3, MOR126-1
          Chromosome Location:
          Chr.11: 51027004 - 51027945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448076_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010997.1 NP_035127.1 1 746 76
          GenBank mRNA AF102530.1 - 1 551 76
          BC099884.1 - 1 745 76
          BC099885.1 - 1 745 76
          BC100678.1 - 1 745 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000558.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr54

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014388 AF102530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014388 BC099884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014388 BC099885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014388 NM_010997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014388 BC100678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014388 AF102530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014388 BC099884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014388 BC099885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014388 NM_010997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014388 BC100678.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117206 AF102530.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117206 BC099884.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117206 BC099885.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117206 NM_010997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117206 BC100678.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791899 - -
          Whole Mouse Genome G4122A A_51_P117206 AF102530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117206 BC099884.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117206 BC099885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117206 NM_010997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117206 BC100678.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09707718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09017069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09600005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09227842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707726 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375796 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375797 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375795 - -
          Affymetrix Mouse Clariom S Array PSR1100004304.mm.1 TC1100000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004304.mm.1 NM_010997.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004304.mm.1 NM_010997.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 783167 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2690494 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249015 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249015 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249015 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0