Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163671.1 | NP_001157143.1 | 3 - 4 | 532 | 122 | |
NM_001163672.1 | NP_001157144.1 | 3 - 4 | 532 | 122 | ||
NM_016961.3 | NP_058657.1 | 3 - 4 | 532 | 122 | ||
NM_207692.2 | NP_997575.2 | 3 - 4 | 532 | 122 | ||
GenBank mRNA | AB005664.1 | - | 2 - 3 | 306 | 122 | |
AF052466.1 | - | 2 - 3 | 268 | 122 | ||
AF052467.1 | - | 2 - 3 | 268 | 122 | ||
AF052468.1 | - | 2 - 3 | 268 | 122 | ||
AF052469.1 | - | 2 - 3 | 268 | 122 | ||
AK031959.1 | - | 3 - 4 | 531 | 122 | ||
AK031974.1 | - | 1 - 2 | 86 | 122 | ||
AK145083.1 | - | 3 - 4 | 507 | 122 | ||
BC028341.1 | - | 3 - 4 | 514 | 4192796 | 122 | |
CX235052.1 | - | 3 - 4 | 515 | 122 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |