Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441062_m1
          See other RAB18 GE Assays ›
          Gene Symbol:
          Rab18
          Gene Name:
          RAB18, member RAS oncogene family
          Gene Aliases:
          AA959686
          Chromosome Location:
          Chr.18: 6765014 - 6791606 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441062_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001278447.1 NP_001265376.1 6 - 7 558 74
          NM_181070.6 NP_851415.1 6 - 7 540 74
          GenBank mRNA AB232612.1 - 6 - 7 442 74
          AK028155.1 - 2 - 3 141 74
          AK045514.1 - 6 - 7 503 74
          AK140901.1 - 6 - 7 508 74
          AK143673.1 - 6 - 7 516 74
          AK146177.1 - 6 - 7 465 74
          AK146397.1 - 6 - 7 465 74
          AK154920.1 - 6 - 7 517 74
          BC056351.1 - 6 - 7 494 6807284 74
          L04966.1 - 6 - 7 446 74
          X80333.1 - 6 - 7 442 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000047.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab18

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          small GTPase mediated signal transduction
          multicellular organism development
          protein transport
          Rab protein signal transduction
          lipid droplet organization
          endoplasmic reticulum tubular network organization
          nucleotide binding
          GTPase activity
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 BC056351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK154920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK140901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK143673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK088760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 BC056351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK154920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK140901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK143673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P327615 AK088760.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418049 AK007078.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418049 AK007251.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418049 BC049568.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418049 NM_026104.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 162723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33894 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 BC056351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK154920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK140901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK143673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK088760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 BC056351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK154920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK140901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK143673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P327615 AK088760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215496 BC056351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215496 L04966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215496 AK146397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215496 AK154920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215496 AK140901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 BC056351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK154920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK140901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK143673.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK088760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 BC056351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK154920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK140901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK143673.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327615 AK088760.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31474606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14043476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474608 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10453709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453711 - -
          Affymetrix HT MG-430 PM Array Plate 1420899_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420900_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10453705 - -
          Mouse Genome 430 2.0 Array 1420900_a_at - -
          Mouse Genome 430 2.0 Array 1420899_PM_at - -
          Mouse Genome 430 2.0 Array 1420899_at - -
          Mouse Genome 430 2.0 Array 1420900_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19330_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19330_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800000304.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000304.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000304.mm.1 NM_181070.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000299.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000299.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000299.mm.1 NM_181070.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000301.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000301.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000301.mm.1 NM_181070.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000302.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000302.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000302.mm.1 NM_181070.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000305.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000305.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000305.mm.1 NM_181070.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000300.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000300.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000300.mm.1 NM_181070.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000303.mm.1 TC1800000047.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000303.mm.1 NM_001278447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000303.mm.1 NM_181070.6 Refseq
          Mouse Expression Set 430 1420899_PM_at - -
          Mouse Expression Set 430 1420900_PM_a_at - -
          Mouse Expression Set 430 1420900_a_at - -
          Mouse Expression Set 430 1420899_at - -
          Murine Genome U74v2 Set 94319_at - -
          Murine Genome U74v2 Set 94319_PM_at - -
          Mouse Genome 430A 2.0 Array 1420899_at - -
          Mouse Genome 430A 2.0 Array 1420899_PM_at - -
          Mouse Genome 430A 2.0 Array 1420900_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1420900_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420899_at - -
          Affymetrix HT Mouse Genome 430A Array 1420900_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420899_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420900_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4590577 - -
          MouseWG-6 v2 Expression BeadChip 106130537 - -
          MouseWG-6 v2 Expression BeadChip 3120575 - -
          Illumina mouse-6 v1.0 expression beadchip 3120575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217153 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217153 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689728 - -
          mouse-6 v1.1 (Illumina) scl0002187.1_532-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002187.1_532-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689728 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217153 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217153 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002187.1_532-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0