Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436213_g1
          See other RAB24 GE Assays ›
          Gene Symbol:
          Rab24
          Gene Name:
          RAB24, member RAS oncogene family
          Gene Aliases:
          6530406O07Rik
          Chromosome Location:
          Chr.13: 55319223 - 55321980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436213_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009000.3 NP_033026.1 2 - 3 511 53
          GenBank mRNA AB232619.1 - 2 - 3 185 53
          AK007602.1 - 2 - 3 474 53
          AK160309.1 - 1 - 2 554 53
          BC019950.1 - 2 - 3 465 4011304 53
          BC028872.1 - 2 - 3 417 4220908 53
          BC054466.1 - 2 - 3 476 6530844 53
          BC055894.1 - 2 - 3 474 3964529 53
          BX526228.1 - 2 - 3 467 53
          Z22819.1 - 2 - 3 185 53

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002052.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab24

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          autophagy
          small GTPase mediated signal transduction
          protein transport
          Rab protein signal transduction
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300012919.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012919.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012919.mm.1 NM_009000.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012917.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012917.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012917.mm.1 NM_009000.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012924.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012924.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012924.mm.1 NM_009000.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012916.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012916.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012916.mm.1 NM_009000.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012920.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012920.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012920.mm.1 NM_009000.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012918.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012918.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012918.mm.1 NM_009000.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012921.mm.1 TC1300002052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012921.mm.1 NM_009000.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012921.mm.1 NM_009000.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 674333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-b98c95889-pjv49:80/100.66.134.79:80.
          git-commit: ef20cda39f797f656ba0530dba66184d0857c864
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0