Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435471_m1
          See other P2RY1 GE Assays ›
          Gene Symbol:
          P2ry1
          Gene Name:
          purinergic receptor P2Y, G-protein coupled 1
          Gene Aliases:
          P2Y1
          Chromosome Location:
          Chr.3: 61002785 - 61008982 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435471_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2ry1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008772.5 NP_032798.1 1 - 2 1820 81
          GenBank mRNA AK036611.1 - 1 - 2 1811 81
          BC138634.1 - 1 - 2 1681 8861654 81
          BC138635.1 - 1 - 2 1681 8861655 81
          U22829.1 - 1 - 2 1194 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000491.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2ry1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          G protein-coupled adenosine receptor signaling pathway
          signal transduction
          G protein-coupled receptor signaling pathway
          adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
          phospholipase C-activating G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          glial cell migration
          response to mechanical stimulus
          regulation of signaling receptor activity
          negative regulation of norepinephrine secretion
          sensory perception of pain
          signal transduction involved in regulation of gene expression
          platelet activation
          positive regulation of inositol trisphosphate biosynthetic process
          G protein-coupled purinergic nucleotide receptor signaling pathway
          eating behavior
          positive regulation of ion transport
          positive regulation of transcription by RNA polymerase II
          positive regulation of hormone secretion
          negative regulation of binding
          positive regulation of penile erection
          positive regulation of ERK1 and ERK2 cascade
          cellular response to organic cyclic compound
          protein localization to plasma membrane
          relaxation of muscle
          blood vessel diameter maintenance
          G protein-coupled ADP receptor activity
          G protein-coupled receptor activity
          ATP binding
          A1 adenosine receptor binding
          ADP binding
          G protein-coupled purinergic nucleotide receptor activity
          G protein-coupled ATP receptor activity
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300003399.mm.1 TC0300000491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003399.mm.1 NM_001282016.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003399.mm.1 NM_008772.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003401.mm.1 TC0300000491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003401.mm.1 NM_001282016.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003401.mm.1 NM_008772.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003398.mm.1 TC0300000491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003398.mm.1 NM_001282016.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003398.mm.1 NM_008772.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003400.mm.1 TC0300000491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003400.mm.1 NM_008772.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 544174 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0