Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433852_g1
          See other GRN GE Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          Pgrn, epithelin
          Chromosome Location:
          Chr.11: 102430322 - 102436809 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433852_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008175.4 NP_032201.2 8 - 9 1055 62
          GenBank mRNA AK013678.1 - 8 - 9 871 62
          AK018744.1 - 8 - 9 896 62
          AK147932.1 - 8 - 9 872 62
          AK149570.1 - 8 - 9 874 62
          AK150125.1 - 8 - 9 870 62
          AK150205.1 - 8 - 9 871 62
          AK150338.1 - 8 - 9 860 62
          AK150420.1 - 8 - 9 859 62
          AK150717.1 - 8 - 9 871 62
          AK150746.1 - 8 - 9 872 62
          AK150881.1 - 8 - 9 872 62
          AK151007.1 - 8 - 9 871 62
          AK151162.1 - 8 - 9 872 62
          AK151343.1 - 8 - 9 871 62
          AK151713.1 - 8 - 9 871 62
          AK151756.1 - 8 - 9 871 62
          AK152048.1 - 8 - 9 871 62
          AK152087.1 - 8 - 9 871 62
          AK152118.1 - 8 - 9 860 62
          AK152213.1 - 8 - 9 860 62
          AK152705.1 - 8 - 9 871 62
          AK152765.1 - 8 - 9 871 62
          AK152944.1 - 8 - 9 860 62
          AK153183.1 - 8 - 9 860 62
          AK153269.1 - 8 - 9 871 62
          AK153289.1 - 8 - 9 871 62
          AK153418.1 - 8 - 9 871 62
          AK153470.1 - 8 - 9 871 62
          AK154492.1 - 8 - 9 871 62
          AK159397.1 - 8 - 9 860 62
          AK159428.1 - 8 - 9 860 62
          AK159557.1 - 8 - 9 871 62
          AK159809.1 - 8 - 9 871 62
          AK160022.1 - 8 - 9 860 62
          AK160033.1 - 8 - 9 871 62
          AK160074.1 - 8 - 9 860 62
          AK211032.1 - 3 - 4 324 62
          AK219696.1 - 3 - 4 334 62
          BC037047.1 - 7 - 8 852 62
          BC049943.1 - 8 - 9 857 62
          BC129849.1 - 8 - 9 855 40109630 62
          BC129850.1 - 8 - 9 855 40109631 62
          M86736.1 - 8 - 9 853 62
          X62321.1 - 7 - 8 836 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grn

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          RNA binding
          cytokine activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100015916.mm.1 TC1100001688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015916.mm.1 NM_008175.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015916.mm.1 NM_008175.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0