Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433851_g1
          See other GRN GE Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          Pgrn, epithelin
          Chromosome Location:
          Chr.11: 102430322 - 102436809 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433851_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008175.4 NP_032201.2 7 - 8 931 92
          GenBank mRNA AK013678.1 - 7 - 8 747 92
          AK018744.1 - 7 - 8 772 92
          AK147932.1 - 7 - 8 748 92
          AK149570.1 - 7 - 8 750 92
          AK150125.1 - 7 - 8 746 92
          AK150205.1 - 7 - 8 747 92
          AK150338.1 - 7 - 8 736 92
          AK150420.1 - 7 - 8 735 92
          AK150717.1 - 7 - 8 747 92
          AK150746.1 - 7 - 8 748 92
          AK150881.1 - 7 - 8 748 92
          AK151007.1 - 7 - 8 747 92
          AK151162.1 - 7 - 8 748 92
          AK151343.1 - 7 - 8 747 92
          AK151713.1 - 7 - 8 747 92
          AK151756.1 - 7 - 8 747 92
          AK152048.1 - 7 - 8 747 92
          AK152087.1 - 7 - 8 747 92
          AK152118.1 - 7 - 8 736 92
          AK152213.1 - 7 - 8 736 92
          AK152705.1 - 7 - 8 747 92
          AK152765.1 - 7 - 8 747 92
          AK152944.1 - 7 - 8 736 92
          AK153183.1 - 7 - 8 736 92
          AK153269.1 - 7 - 8 747 92
          AK153289.1 - 7 - 8 747 92
          AK153418.1 - 7 - 8 747 92
          AK153470.1 - 7 - 8 747 92
          AK154492.1 - 7 - 8 747 92
          AK159397.1 - 7 - 8 736 92
          AK159428.1 - 7 - 8 736 92
          AK159557.1 - 7 - 8 747 92
          AK159809.1 - 7 - 8 747 92
          AK160033.1 - 7 - 8 747 92
          AK160074.1 - 7 - 8 736 92
          AK211032.1 - 2 - 3 200 92
          AK219696.1 - 2 - 3 210 92
          BC037047.1 - 6 - 7 728 92
          BC049943.1 - 7 - 8 733 92
          BC129849.1 - 7 - 8 731 40109630 92
          BC129850.1 - 7 - 8 731 40109631 92
          X62321.1 - 6 - 7 712 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grn

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          RNA binding
          cytokine activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100015908.mm.1 TC1100001688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015908.mm.1 NM_008175.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015908.mm.1 NM_008175.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015909.mm.1 TC1100001688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015909.mm.1 NM_008175.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015909.mm.1 NM_008175.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015913.mm.1 TC1100001688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015913.mm.1 NM_008175.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015913.mm.1 NM_008175.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6dd4f99df8-cwvvz:80/100.66.134.79:80.
          git-commit: 33aac1f801e405ffbac6f281a4ca9f2baedc1304
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0