Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433849_g1
          See other GRN GE Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          Pgrn, epithelin
          Chromosome Location:
          Chr.11: 102430322 - 102436809 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433849_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008175.4 NP_032201.2 5 - 6 685 77
          GenBank mRNA AK013678.1 - 5 - 6 501 77
          AK018744.1 - 5 - 6 526 77
          AK147932.1 - 5 - 6 501 77
          AK149570.1 - 5 - 6 504 77
          AK150125.1 - 5 - 6 500 77
          AK150205.1 - 5 - 6 501 77
          AK150338.1 - 5 - 6 490 77
          AK150420.1 - 5 - 6 489 77
          AK150717.1 - 5 - 6 501 77
          AK150746.1 - 5 - 6 502 77
          AK150881.1 - 5 - 6 502 77
          AK151007.1 - 5 - 6 501 77
          AK151162.1 - 5 - 6 502 77
          AK151343.1 - 5 - 6 501 77
          AK151713.1 - 5 - 6 501 77
          AK151756.1 - 5 - 6 501 77
          AK152048.1 - 5 - 6 501 77
          AK152087.1 - 5 - 6 501 77
          AK152118.1 - 5 - 6 490 77
          AK152213.1 - 5 - 6 490 77
          AK152705.1 - 5 - 6 501 77
          AK152765.1 - 5 - 6 501 77
          AK152944.1 - 5 - 6 490 77
          AK153183.1 - 5 - 6 490 77
          AK153269.1 - 5 - 6 501 77
          AK153289.1 - 5 - 6 501 77
          AK153418.1 - 5 - 6 501 77
          AK154492.1 - 5 - 6 501 77
          AK159397.1 - 5 - 6 490 77
          AK159428.1 - 5 - 6 490 77
          AK159557.1 - 5 - 6 501 77
          AK159809.1 - 5 - 6 501 77
          AK160022.1 - 5 - 6 490 77
          AK160033.1 - 5 - 6 501 77
          AK160074.1 - 5 - 6 490 77
          BC037047.1 - 4 - 5 482 77
          BC049943.1 - 5 - 6 487 77
          BC129849.1 - 5 - 6 485 40109630 77
          BC129850.1 - 5 - 6 485 40109631 77
          M86736.1 - 5 - 6 483 77
          X62321.1 - 4 - 5 466 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grn

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          RNA binding
          cytokine activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100015906.mm.1 TC1100001688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015906.mm.1 NM_008175.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015906.mm.1 NM_008175.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-hbrf6:80/100.66.134.79:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0