Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs99999029_m1
          See other IL1B GE Assays ›
          Gene Symbol:
          IL1B
          Gene Name:
          interleukin 1 beta
          Gene Aliases:
          IL-1, IL1-BETA, IL1F2
          Chromosome Location:
          Chr.2: 112829758 - 112836842 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs99999029_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000576.2 NP_000567.1 3 - 4 188 97
          XM_017003988.1 XP_016859477.1 2 - 3 189 97
          GenBank mRNA AK313983.1 - 2 - 3 651 97
          BC008678.1 - 3 - 4 188 3875593 97
          BT007213.1 - 2 - 3 101 97
          CR407679.1 - 2 - 3 101 97
          K02770.1 - 3 - 4 187 97
          M15330.1 - 3 - 4 187 97
          X02532.1 - 3 - 4 178 97
          X56087.1 - 3 - 4 158 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013916.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002219.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017003988.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL1B

          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          activation of MAPK activity
          fever generation
          response to hypoxia
          positive regulation of protein phosphorylation
          chronic inflammatory response to antigenic stimulus
          positive regulation of T cell mediated immunity
          purine nucleobase metabolic process
          apoptotic process
          inflammatory response
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          embryo implantation
          aging
          memory
          estrogen metabolic process
          negative regulation of cell proliferation
          glycoprotein metabolic process
          response to heat
          response to ozone
          response to gamma radiation
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          negative regulation of glucose transport
          negative regulation of glutamate secretion
          smooth muscle adaptation
          cytokine-mediated signaling pathway
          pentacyclic triterpenoid metabolic process
          hyaluronan biosynthetic process
          neutrophil chemotaxis
          polyketide metabolic process
          ovulation
          sequestering of triglyceride
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of fever generation
          lipopolysaccharide-mediated signaling pathway
          positive regulation of prostaglandin secretion
          response to estradiol
          interleukin-1 beta production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of immature T cell proliferation in thymus
          positive regulation of histone phosphorylation
          response to ATP
          response to vitamin D
          response to L-ascorbic acid
          positive regulation of heterotypic cell-cell adhesion
          positive regulation of histone acetylation
          social behavior
          ectopic germ cell programmed cell death
          positive regulation of myosin light chain kinase activity
          response to stilbenoid
          cellular response to drug
          response to diuretic
          response to statin
          wound healing
          positive regulation of T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          negative regulation of MAP kinase activity
          response to peptide hormone
          protein kinase B signaling
          positive regulation of JUN kinase activity
          positive regulation of chemokine biosynthetic process
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          negative regulation of neuron differentiation
          positive regulation of angiogenesis
          negative regulation of lipid metabolic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          negative regulation of insulin receptor signaling pathway
          positive regulation of protein export from nucleus
          positive regulation of astrocyte differentiation
          positive regulation of phagocytosis
          regulation of insulin secretion
          negative regulation of lipid catabolic process
          positive regulation of lipid catabolic process
          regulation of nitric-oxide synthase activity
          positive regulation of membrane protein ectodomain proteolysis
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of cell division
          positive regulation of cell adhesion molecule production
          positive regulation of calcidiol 1-monooxygenase activity
          negative regulation of adiponectin secretion
          positive regulation of ERK1 and ERK2 cascade
          monocyte aggregation
          cellular response to antibiotic
          cellular response to mechanical stimulus
          cellular response to organic substance
          cellular response to glucose stimulus
          cellular response to fatty acid
          cellular response to organic cyclic compound
          cellular response to methotrexate
          response to dexamethasone
          positive regulation of monocyte chemotactic protein-1 production
          positive regulation of neutrophil chemotaxis
          extrinsic apoptotic signaling pathway in absence of ligand
          regulation of establishment of endothelial barrier
          negative regulation of branching morphogenesis of a nerve
          negative regulation of neural precursor cell proliferation
          positive regulation of interleukin-6 secretion
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          cytokine activity
          interleukin-1 receptor binding
          protein domain specific binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000263341.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000416750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000416750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000416750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000416750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000416750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000418817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000432018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000432018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000432018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000432018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000432018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000432018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000477398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000477398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000477398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000477398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000487639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000491056.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000496280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000496280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000496280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000496280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000496280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 ENST00000496280.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013916.hg.1 NM_000576.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7dc8547fcc-q6vt2:80/100.66.131.252:80.
          git-commit: cbebc3e589bdd6cb3539df712021cd796c57f930
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0