Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs99999008_m1
          See other MDM2 GE Assays ›
          Gene Symbol:
          MDM2
          Gene Name:
          MDM2 proto-oncogene
          Gene Aliases:
          ACTFS, HDMX, hdm2
          Chromosome Location:
          Chr.12: 68808149 - 68845544 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs99999008_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MDM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145339.2 NP_001138811.1 2 346 73
          NM_002392.5 NP_002383.2 2 346 73
          XM_006719400.3 XP_006719463.1 2 369 73
          GenBank mRNA AF092843.1 - 2 54 73
          AF092844.1 - 2 54 73
          AF092845.1 - 2 54 73
          AK290341.1 - 2 296 73
          BE296905.1 - 2 311 73
          HY174841.1 - 2 348 73
          M92424.1 - 2 333 73
          Z12020.1 - 2 333 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008116.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF092843.1,AF092844.1,AF092845.1,AK290341.1,M92424.1,XM_006719400.3,Z12020.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Homo sapiens Human p53 Signaling HUMAN DNA REPAIR MECHANISM HUMAN MOLECULAR MECHANISMS OF CANCER HUMAN P53 MEDIATED APOPTOSIS HUMAN CELLULAR APOPTOSIS PATHWAY HUMAN PANCREATIC ADENOCARCINOMA Inventoried Human_p53_Signaling Human_Pancreatic_Adenocarcinoma Human_Molecular_Mechanisms_of_Cancer Human_p53_Mediated_Apoptosis Human_Cellular_Apoptosis_Pathway Human_DNA_Repair_Mechanism Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MDM2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          blood vessel remodeling
          regulation of heart rate
          atrioventricular valve morphogenesis
          endocardial cushion morphogenesis
          ventricular septum development
          atrial septum development
          protein complex assembly
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          traversing start control point of mitotic cell cycle
          positive regulation of cell proliferation
          response to toxic substance
          response to carbohydrate
          response to iron ion
          positive regulation of gene expression
          negative regulation of protein processing
          viral process
          protein ubiquitination
          protein sumoylation
          peptidyl-lysine modification
          protein destabilization
          response to magnesium ion
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein localization to nucleus
          regulation of protein catabolic process
          response to cocaine
          response to drug
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to morphine
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          establishment of protein localization
          response to ether
          negative regulation of transcription, DNA-templated
          positive regulation of mitotic cell cycle
          response to antibiotic
          positive regulation of protein export from nucleus
          response to steroid hormone
          cardiac septum morphogenesis
          cellular response to hydrogen peroxide
          negative regulation of cell cycle arrest
          cellular response to antibiotic
          cellular response to vitamin B1
          cellular response to alkaloid
          cellular response to growth factor stimulus
          cellular response to peptide hormone stimulus
          cellular response to estrogen stimulus
          cellular response to hypoxia
          cellular response to UV-C
          regulation of signal transduction by p53 class mediator
          negative regulation of signal transduction by p53 class mediator
          p53 binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          SUMO transferase activity
          enzyme binding
          ubiquitin protein ligase binding
          identical protein binding
          peroxisome proliferator activated receptor binding
          ubiquitin protein ligase activity
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000258148.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000258149.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000299252.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000311420.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000348801.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000350057.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000356290.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000360430.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393410.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393412.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393413.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393415.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393417.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000428863.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000462284.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000478070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000481186.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000493419.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000496959.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000517852.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000523991.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000536089.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000537182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000539479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000540352.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000542502.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000543323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000544561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000545204.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000546048.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001145337.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001145339.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001145340.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001278462.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_002392.5 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000258148.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000258149.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000299252.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000311420.13 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000348801.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000350057.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000356290.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000360430.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393410.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393412.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393413.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393415.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000393417.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000428863.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000462284.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000478070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000481186.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000493419.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000496959.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000517852.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000523991.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000536089.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000537182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000539479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000540352.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000542502.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000543323.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000544561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000545204.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 ENST00000546048.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001145337.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001145339.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001145340.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_001278462.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 NM_002392.5 Refseq
          Affymetrix Human Clariom D Assay PSR1200159431.hg.1 TC1200008116.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0