Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07319937_m1
          See other SH3BP5-AS1 GE Assays ›
          Gene Symbol:
          SH3BP5-AS1
          Gene Name:
          SH3BP5 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 15254184 - 15264498 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07319937_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SH3BP5-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046084.1 2 - 3 5371 74
          GenBank mRNA AL133111.1 - 2 - 3 5372 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM995581.1 - 1 - 2 212 Chr. 3 - 15260228 - 15264493 [-] on Build GRCh38 74
          GenBankmRNA AI039273.1 - 1 - 2 212 Chr. 3 - 15260369 - 15264500 [-] on Build GRCh38 74
          GenBankmRNA AI290027.1 - 1 - 2 172 Chr. 3 - 15260387 - 15264465 [-] on Build GRCh38 74
          GenBankmRNA CA436406.1 - 1 - 2 221 Chr. 3 - 15260465 - 15264500 [-] on Build GRCh38 74
          GenBankmRNA AW770980.1 - 1 - 2 195 Chr. 3 - 15260276 - 15264485 [-] on Build GRCh38 74
          GenBankmRNA BQ581542.1 - 1 - 2 231 Chr. 3 - 15260276 - 15264500 [-] on Build GRCh38 74
          GenBankmRNA BI496472.1 - 1 - 2 199 Chr. 3 - 15260386 - 15264481 [+] on Build GRCh38 74
          GenBankmRNA AW444814.1 - 1 - 2 221 Chr. 3 - 15260396 - 15264500 [-] on Build GRCh38 74
          GenBankmRNA BM719762.1 - 1 - 2 394 Chr. 3 - 15260188 - 15264500 [+] on Build GRCh38 74
          GenBankmRNA BF939098.1 - 1 - 2 199 Chr. 3 - 15260280 - 15264492 [-] on Build GRCh38 74
          GenBankmRNA CK903231.1 - 1 - 2 187 Chr. 3 - 15259492 - 15264480 [-] on Build GRCh38 74
          GenBankmRNA AI754162.1 - 1 - 2 Chr. 3 - 15260327 - 15264493 [-] on Build GRCh38 74
          GenBankmRNA AA233894.1 - 1 - 2 195 Chr. 3 - 15260384 - 15264488 [-] on Build GRCh38 74
          GenBankmRNA BG057676.1 - 1 - 2 196 Chr. 3 - 15260228 - 15264489 [-] on Build GRCh38 74
          GenBankmRNA BF527961.1 - 65 Chr. - - [-] on Build 74
          GenBankmRNA BM995561.1 - 1 - 2 212 Chr. 3 - 15260179 - 15264493 [-] on Build GRCh38 74
          GenBankmRNA AA007601.1 - 1 - 2 205 Chr. 3 - 15260265 - 15264498 [-] on Build GRCh38 74
          GenBankmRNA AW444539.1 - 1 - 2 211 Chr. 3 - 15260314 - 15264493 [-] on Build GRCh38 74
          GenBankmRNA AI381661.1 - 1 - 2 206 Chr. 3 - 15260437 - 15264498 [-] on Build GRCh38 74
          GenBankmRNA BE673690.1 - 1 - 2 199 Chr. 3 - 15260280 - 15264492 [-] on Build GRCh38 74
          GenBankmRNA N66918.1 - 1 - 2 203 Chr. 3 - 15260343 - 15264497 [-] on Build GRCh38 74
          GenBankmRNA BE218198.1 - 1 - 2 195 Chr. 3 - 15260256 - 15264488 [-] on Build GRCh38 74
          GenBankmRNA DB364531.1 - 1 - 2 196 Chr. 3 - 15260241 - 15264489 [-] on Build GRCh38 74
          GenBankmRNA BM684117.1 - 1 - 2 222 Chr. 3 - 15260181 - 15264500 [-] on Build GRCh38 74
          GenBankmRNA AW028372.1 - 1 - 2 205 Chr. 3 - 15260435 - 15264498 [-] on Build GRCh38 74
          GenBankmRNA AW072432.1 - 1 - 2 197 Chr. 3 - 15260256 - 15264490 [-] on Build GRCh38 74
          GenBankmRNA AA761364.1 - 1 - 2 205 Chr. 3 - 15260267 - 15264493 [-] on Build GRCh38 74
          GenBankmRNA AW236970.1 - 1 - 2 198 Chr. 3 - 15260235 - 15264491 [-] on Build GRCh38 74
          GenBankmRNA BE466779.1 - 1 - 2 198 Chr. 3 - 15260342 - 15264491 [-] on Build GRCh38 74
          GenBankmRNA AI681330.1 - 1 - 2 195 Chr. 3 - 15260335 - 15264488 [-] on Build GRCh38 74
          GenBankmRNA AI242320.1 - 1 - 2 194 Chr. 3 - 15260366 - 15264487 [-] on Build GRCh38 74
          GenBankmRNA AA704194.1 - 1 - 2 197 Chr. 3 - 15260533 - 15264490 [-] on Build GRCh38 74
          GenBankmRNA AW044592.1 - 1 - 2 203 Chr. 3 - 15260350 - 15264495 [-] on Build GRCh38 74
          GenBankmRNA AI123416.1 - 1 - 2 204 Chr. 3 - 15260217 - 15264497 [-] on Build GRCh38 74
          GenBankmRNA AI688471.1 - 1 - 2 201 Chr. 3 - 15260299 - 15264493 [-] on Build GRCh38 74
          GenBankmRNA AI061466.1 - 1 - 2 207 Chr. 3 - 15260334 - 15264493 [-] on Build GRCh38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream calpain 7 CAPN7 CALPAIN7,
          PALBH
          -5.7kb Sense Chr. 3 - 15206226 - 15252916 on NCBI Build GRCh38
          Downstream methyltransferase like 6 METTL6 116.8kb Anti-sense Chr. 3 - 15381275 - 15427580 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0