Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06636186_s1
          See other CACNA1C GE Assays ›
          Gene Symbol:
          CACNA1C
          Gene Name:
          calcium voltage-gated channel subunit alpha1 C
          Gene Aliases:
          CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
          Chromosome Location:
          Chr.12: 1969677 - 2697949 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06636186_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CACNA1C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000719.6 NP_000710.5 10 1762 58
          NM_001129827.1 NP_001123299.1 10 1762 58
          NM_001129829.1 NP_001123301.1 10 1762 58
          NM_001129830.2 NP_001123302.2 10 1727 58
          NM_001129831.1 NP_001123303.1 10 1762 58
          NM_001129832.1 NP_001123304.1 10 1762 58
          NM_001129833.1 NP_001123305.1 10 1762 58
          NM_001129834.1 NP_001123306.1 10 1762 58
          NM_001129835.1 NP_001123307.1 10 1762 58
          NM_001129836.1 NP_001123308.1 10 1762 58
          NM_001129837.1 NP_001123309.1 10 1762 58
          NM_001129838.1 NP_001123310.1 10 1762 58
          NM_001129839.1 NP_001123311.1 10 1762 58
          NM_001129840.1 NP_001123312.1 10 1762 58
          NM_001129841.1 NP_001123313.1 10 1762 58
          NM_001129842.1 NP_001123314.1 10 1762 58
          NM_001129843.1 NP_001123315.1 10 1762 58
          NM_001129844.1 NP_001123316.1 10 1753 58
          NM_001129846.1 NP_001123318.1 10 1762 58
          NM_001167623.1 NP_001161095.1 10 1762 58
          NM_001167624.2 NP_001161096.2 10 1727 58
          NM_001167625.1 NP_001161097.1 10 1762 58
          NM_199460.3 NP_955630.3 10 1727 58
          XM_006719017.2 XP_006719080.1 10 1816 58
          XM_011521020.2 XP_011519322.1 11 1891 58
          XM_011521023.2 XP_011519325.1 11 2076 58
          XM_017019926.1 XP_016875415.1 12 3078 58
          XM_017019927.1 XP_016875416.1 12 3078 58
          XM_017019928.1 XP_016875417.1 12 3078 58
          XM_017019929.1 XP_016875418.1 12 3078 58
          XM_017019930.1 XP_016875419.1 12 3078 58
          XM_017019931.1 XP_016875420.1 12 3078 58
          XM_017019932.1 XP_016875421.1 11 3003 58
          XM_017019933.1 XP_016875422.1 11 3003 58
          XM_017019934.1 XP_016875423.1 12 3078 58
          XM_017019935.1 XP_016875424.1 12 3078 58
          XM_017019936.1 XP_016875425.1 11 3003 58
          XM_017019937.1 XP_016875426.1 11 3003 58
          XM_017019938.1 XP_016875427.1 11 3003 58
          XM_017019939.1 XP_016875428.1 11 3003 58
          XM_017019940.1 XP_016875429.1 11 3003 58
          XM_017019941.1 XP_016875430.1 11 3003 58
          XM_017019942.1 XP_016875431.1 11 3003 58
          XM_017019943.1 XP_016875432.1 11 3003 58
          XM_017019944.1 XP_016875433.1 11 3003 58
          XM_017019945.1 XP_016875434.1 11 3003 58
          XM_017019946.1 XP_016875435.1 11 3003 58
          XM_017019947.1 XP_016875436.1 11 3003 58
          XM_017019948.1 XP_016875437.1 11 3003 58
          XM_017019949.1 XP_016875438.1 11 3003 58
          XM_017019950.1 XP_016875439.1 11 3003 58
          XM_017019951.1 XP_016875440.1 11 2994 58
          XM_017019952.1 XP_016875441.1 11 3003 58
          XM_017019953.1 XP_016875442.1 10 1816 58
          XM_017019954.1 XP_016875443.1 10 1816 58
          XM_017019955.1 XP_016875444.1 12 3078 58
          GenBank mRNA AB209016.1 - 8 1197 58
          AF465484.1 - 10 1714 58
          AJ224873.1 - 10 1449 58
          AJ536834.1 - 3 195 58
          AJ537510.1 - 2 120 58
          AY562395.1 - 4 411 58
          AY562396.1 - 3 336 58
          AY830711.1 - 10 1449 58
          AY830712.1 - 10 1449 58
          AY830713.1 - 10 1449 58
          BC146846.1 - 10 1568 9056774 58
          L29529.1 - 9 1381 58
          L29534.1 - 9 1375 58
          L29536.1 - 9 1375 58
          Z34809.1 - 10 1449 58
          Z34810.1 - 10 1449 58
          Z34811.1 - 10 1449 58
          Z34812.1 - 10 1449 58
          Z34813.1 - 10 1449 58
          Z34814.1 - 10 1449 58
          Z34815.1 - 10 1449 58
          Z34816.1 - 10 1449 58
          Z34817.1 - 10 1449 58
          Z34818.1 - 10 1449 58
          Z34819.1 - 10 1449 58
          Z34820.1 - 10 1449 58
          Z34821.1 - 10 1449 58
          Z34822.1 - 10 1449 58
          Z74996.1 - 10 1449 58

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CACNA1C

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system development
          positive regulation of cytosolic calcium ion concentration
          heart development
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          embryonic forelimb morphogenesis
          calcium-mediated signaling using extracellular calcium source
          camera-type eye development
          regulation of insulin secretion
          calcium ion transport into cytosol
          cardiac conduction
          calcium ion transmembrane transport via high voltage-gated calcium channel
          calcium ion transmembrane transport
          cardiac muscle cell action potential involved in contraction
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during AV node cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          regulation of heart rate by cardiac conduction
          regulation of ventricular cardiac muscle cell action potential
          membrane depolarization during atrial cardiac muscle cell action potential
          voltage-gated calcium channel activity
          protein binding
          calmodulin binding
          high voltage-gated calcium channel activity
          metal ion binding
          alpha-actinin binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved in AV node cell action potential

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vq47s:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0