Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05640888_g1
See other ATXN2L GE Assays ›
Gene Symbol:
ATXN2L
Gene Name:
ataxin 2 like
Gene Aliases:
A2D, A2LG, A2LP, A2RP
Chromosome Location:
Chr.16: 28823048 - 28837237 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs05640888_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ATXN2L
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001308230.1
NP_001295159.1
23
3717
95
NM_007245.3
NP_009176.2
22
4250
95
NM_017492.3
NP_059867.3
23
3699
95
NM_145714.2
NP_663760.1
23
3676
95
NM_148414.2
NP_680780.1
24
3864
95
NM_148415.2
NP_680781.1
23
3645
95
NM_148416.2
NP_680782.1
23
3622
95
XM_005255061.1
XP_005255118.1
23
4122
95
XM_005255062.1
XP_005255119.1
24
3880
95
XM_005255064.1
XP_005255121.1
23
4119
95
XM_005255065.1
XP_005255122.1
23
4104
95
XM_005255066.1
XP_005255123.1
23
4101
95
XM_005255067.1
XP_005255124.1
23
4068
95
XM_005255068.1
XP_005255125.1
24
3826
95
XM_005255069.1
XP_005255126.1
22
4271
95
XM_005255070.1
XP_005255127.1
23
3720
95
XM_005255071.1
XP_005255128.1
23
3697
95
XM_005255074.1
XP_005255131.1
23
3666
95
XM_005255075.1
XP_005255132.1
23
3643
95
XM_005255076.1
XP_005255133.1
23
3663
95
XM_005255077.1
XP_005255134.1
23
4005
95
XM_006721007.1
XP_006721070.1
22
4050
95
XM_006721008.1
XP_006721071.1
22
4032
95
XM_006721009.1
XP_006721072.1
23
3999
95
XM_006721011.1
XP_006721074.1
23
3945
95
XM_006721012.3
XP_006721075.1
24
3703
95
XM_011545719.2
XP_011544021.1
24
3885
95
XM_011545722.1
XP_011544024.1
23
3822
95
XM_017022891.1
XP_016878380.1
23
3987
95
XM_017022892.1
XP_016878381.1
23
3804
95
GenBank mRNA
AF034373.1
-
23
4494
95
AF070605.1
-
8
1774
95
AJ317970.1
-
22
4200
95
AJ317971.2
-
23
3632
95
AJ317972.2
-
24
3820
96
AJ317973.2
-
23
3601
95
AJ317974.2
-
23
3578
95
AK289981.1
-
23
3622
95
AL137587.1
-
2
465
95
AY188334.1
-
25
4092
95
AY188335.1
-
25
3740
95
AY188336.1
-
24
3832
95
AY188337.1
-
24
3800
95
AY188338.1
-
24
4316
95
BC010239.2
-
7
2016
95
BC068012.1
-
23
4108
95
BC082760.1
-
23
3495
5812230
95
U70671.1
-
10
1046
95
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
3' Most
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ATXN2L
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytoplasmic mRNA processing body assembly
stress granule assembly
cell-cell adhesion
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-57bdf9945f-4p2c8:80/100.66.134.79:80.
git-commit: baa7f4fdc4fe37bd11ed6fe4edff1157ae59fd06
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0