Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000622195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000622195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009766.hg.1 |
ENST00000622195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000412295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000622195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000622195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009766.hg.1 |
ENST00000622195.1 |
Ensembl |