Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05047224_s1
          See other TP53 GE Assays ›
          Gene Symbol:
          TP53
          Gene Name:
          tumor protein p53
          Gene Aliases:
          BCC7, LFS1, P53, TRP53
          Chromosome Location:
          Chr.17: 7668402 - 7687550 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05047224_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TP53
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000546.5 NP_000537.3 11 127
          NM_001126112.2 NP_001119584.1 11 127
          NM_001126113.2 NP_001119585.1 12 127
          NM_001126114.2 NP_001119586.1 12 127
          NM_001126115.1 NP_001119587.1 7 127
          NM_001126116.1 NP_001119588.1 8 127
          NM_001126117.1 NP_001119589.1 8 127
          NM_001126118.1 NP_001119590.1 10 127
          NM_001276695.1 NP_001263624.1 12 127
          NM_001276696.1 NP_001263625.1 12 127
          NM_001276697.1 NP_001263626.1 7 127
          NM_001276698.1 NP_001263627.1 8 127
          NM_001276699.1 NP_001263628.1 8 127
          NM_001276760.1 NP_001263689.1 11 127
          NM_001276761.1 NP_001263690.1 11 127
          GenBank mRNA AB082923.1 - 11 127
          AF052180.1 - 1 127
          AF307851.1 - 11 127
          AK223026.1 - 5 127
          AK297462.1 - 12 127
          AK297927.1 - 12 127
          BC003596.1 - 11 3544714 127
          DQ186648.1 - 12 1816 130
          DQ186649.1 - 12 1743 130
          DQ186650.1 - 7 1429 130
          DQ186651.1 - 8 1562 130
          DQ186652.1 - 8 1489 130
          DQ191317.1 - 11 1740 130
          DQ286964.1 - 11 1685 130
          FJ207420.1 - 11 1686 130
          JN214348.1 - 10 127
          JN900492.1 - 10 127
          X01405.1 - 8 127

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TP53

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          base-excision repair
          nucleotide-excision repair
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          ER overload response
          Ras protein signal transduction
          multicellular organism development
          cell aging
          protein localization
          cell population proliferation
          negative regulation of cell population proliferation
          determination of adult lifespan
          response to X-ray
          response to gamma radiation
          positive regulation of gene expression
          viral process
          protein sumoylation
          cell differentiation
          negative regulation of cell growth
          DNA damage response, signal transduction by p53 class mediator
          positive regulation of histone deacetylation
          chromatin assembly
          mitotic G1 DNA damage checkpoint signaling
          positive regulation of protein oligomerization
          cellular response to UV
          cellular response to glucose starvation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          entrainment of circadian clock by photoperiod
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of neuron apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          response to antibiotic
          positive regulation of protein export from nucleus
          regulation of mitochondrial membrane permeability
          negative regulation of fibroblast proliferation
          circadian behavior
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of helicase activity
          protein tetramerization
          negative regulation of telomerase activity
          protein-containing complex assembly
          positive regulation of thymocyte apoptotic process
          cellular response to hypoxia
          cellular response to xenobiotic stimulus
          cellular response to ionizing radiation
          intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of release of cytochrome c from mitochondria
          replicative senescence
          oxidative stress-induced premature senescence
          intrinsic apoptotic signaling pathway
          oligodendrocyte apoptotic process
          positive regulation of execution phase of apoptosis
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          regulation of cell cycle G2/M phase transition
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          positive regulation of reactive oxygen species metabolic process
          positive regulation of intrinsic apoptotic signaling pathway
          transcription cis-regulatory region binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          core promoter sequence-specific DNA binding
          DNA-binding transcription activator activity, RNA polymerase II-specific
          protease binding
          p53 binding
          DNA binding
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          copper ion binding
          protein binding
          ATP binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          receptor tyrosine kinase binding
          ubiquitin protein ligase binding
          histone acetyltransferase binding
          identical protein binding
          sequence-specific DNA binding
          protein self-association
          protein heterodimerization activity
          protein N-terminus binding
          chaperone binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0