Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05033232_s1
See other TRPM3 GE Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs05033232_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_011519035.2
XP_011517337.1
27
10848
94
XM_011519036.2
XP_011517338.1
27
13773
94
XM_011519037.2
XP_011517339.1
26
13729
94
XM_011519038.2
XP_011517340.1
26
13720
94
XM_011519039.2
XP_011517341.1
25
13691
94
XM_011519040.2
XP_011517342.1
26
10779
94
XM_011519041.2
XP_011517343.1
26
13681
94
XM_011519042.2
XP_011517344.1
25
10749
94
XM_011519043.2
XP_011517345.1
25
10776
94
XM_011519044.2
XP_011517346.1
25
13652
94
XM_011519045.2
XP_011517347.1
24
10707
94
XM_011519046.2
XP_011517348.1
24
12410
94
XM_011519047.2
XP_011517349.1
26
10740
94
XM_017015141.1
XP_016870630.1
26
10818
94
XM_017015142.1
XP_016870631.1
26
10812
94
XM_017015143.1
XP_016870632.1
25
10782
94
XM_017015144.1
XP_016870633.1
27
10614
94
XM_017015145.1
XP_016870634.1
27
11292
94
XM_017015146.1
XP_016870635.1
27
10690
94
XM_017015147.1
XP_016870636.1
27
12725
94
XM_017015148.1
XP_016870637.1
28
9178
94
XM_017015149.1
XP_016870638.1
27
9148
94
XM_017015150.1
XP_016870639.1
27
9142
94
XM_017015151.1
XP_016870640.1
27
9121
94
XM_017015152.1
XP_016870641.1
28
10856
94
XM_017015153.1
XP_016870642.1
26
9077
94
XM_017015154.1
XP_016870643.1
27
9294
94
XM_017015155.1
XP_016870644.1
27
10778
94
XM_017015156.1
XP_016870645.1
23
10146
94
XM_017015157.1
XP_016870646.1
25
10710
94
XM_017015159.1
XP_016870648.1
27
8869
94
XM_017015160.1
XP_016870649.1
26
8923
94
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
3' Most
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0